Skip to Content
Merck
All Photos(1)

Key Documents

51844

Supelco

Multielement standard solution 4 for ICP

TraceCERT®, in 10% nitric acid

Synonym(s):

Multi-element reference solution 4 for ICP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

quality

in 10% nitric acid

composition

Al, 40 mg/L
As, 40 mg/L
B, 100 mg/L
Ba, 40 mg/L
Be, 10 mg/L
Cd, 10 mg/L
Co, 10 mg/L
Cr, 20 mg/L
Cu, 20 mg/L
Fe, 100 mg/L
Mn, 10 mg/L
Ni, 20 mg/L
Pb, 40 mg/L
Se, 100 mg/L
Tl, 100 mg/L
V, 40 mg/L
Zn, 100 mg/L

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Multielement standard solution 4 can be used as a calibration standard in the determination of heavy metals concentration in noodle samples using inductively coupled plasma optical emission spectrometry system (ICP-OES).[1]

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Analysis Note

In 10% HNO3:
  • 40 mg/L: Al, As, Ba, Pb, V
  • 10 mg/L: Be, Cd, Co, Mn
  • 100 mg/L: B, Fe, Se, Tl, Zn
  • 20 mg/L: Cr, Cu, Ni

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behrouz Tajdar-Oranj et al.
Environmental science and pollution research international, 25(31), 30928-30937 (2018-09-05)
In the current study, the concentration of heavy metals including lead (Pb), chromium (Cr), cadmium (Cd), and aluminum (Al) in commonly instant noodles consumed in Iran (either imported from other countries or produced in Iran) was investigated by acid digestion
Anti-inflammatory activity of linalool and linalyl acetate constituents of essential oils
Peana.T.A, et al.
Phytomedicine, 9, 721-726 (2002)
Hassan Nasirian et al.
Environmental monitoring and assessment, 188(2), 107-107 (2016-01-23)
The Shadegan and Hawr Al Azim wetlands are important natural resources in southwestern Iran, yet relatively little work has been done to assess ecosystem health of the wetlands. Bed sediment from both wetlands was sampled in individual months between October
Reza Shokoohi et al.
Ecotoxicology and environmental safety, 211, 111939-111939 (2021-01-22)
It has been documented that arsenic has a potential risk to human health and identified as a risk factor for hearing impairment. However, there are few studies that confirm the ototoxic effect of arsenic, especially on the human auditory system.
Linalool modifies the nicotinic receptor?ion channel kinetics at the mouse neuromuscular junction
Re.L, et al.
Pharmacological Research, 42, 177-181 (2000)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the elemental composition and respective concentration of 51844-100 ML multi element std. Solution 4 for ICP?

    1 answer
    1. The elemental composition and respective concentrations for this product are located above, in the 'PROPERTIES' section, under composition. The lot specific certified values are reported in the product Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sial/51844#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service