Skip to Content
Merck
All Photos(3)

Key Documents

1.07021

Supelco

Acetone

for analysis EMPARTA® ACS

Synonym(s):

Dimethyl ketone, Propanone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-662-2
NACRES:
NB.21
grade:
ACS reagent
reagent grade
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

grade

ACS reagent
reagent grade

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

description

for analytical purposes
for extraction

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

465 °C (DIN 51794)

potency

5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

expl. lim.

13.2 %

dilution

(for routine analytical applications)

impurities

≤0.0003% Substances reducing potassium permanganate (as O)
≤0.0003 meq/g Titrable acid
≤0.0006 meq/g Titrable base
≤0.002% Aldehydes (as formaldehyde)
≤0.05% 2-Propanol (GC)
≤0.05% Methanol (GC)
≤0.5% Water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

capacity

1000  g/kg water absorbing capacity

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMD Millipore offers EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Packaging

1.07021.0521: 500ML, Minimum order quantity: 20
1.07021.2521: 2.5L, Minimum order quantity: 4

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Solubility in water: conforms
Color: ≤ 10 Hazen
Titrable acid: ≤ 0.0003 meq/g
Titrable base: ≤ 0.0006 meq/g
Methanol (GC): ≤ 0.05 %
2-Propanol (GC): ≤ 0.05 %
Aldehydes (as formaldehyde): ≤ 0.002 %
Substances reducing potassium permanganate (as O): ≤ 0.0003 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.5 %

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service