Skip to Content
Merck
All Photos(1)

Key Documents

1.04403

Supelco

Mercury

for analysis and for polarography EMSURE®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141726
NACRES:
NA.21
form:
liquid

Quality Level

form

liquid

General description

Mercury (quicksilver) is extensively used in the manufacture of mercuric (I) and mercuric (II) compounds because it exists in two oxidation states +1 and +2. It is also used in the preparation of organomercury compounds and coordination complexes of mercury. Mercury(II) salts are used in the preparation of industrially important chemical reagents such as Millon′s base, Nessler′s reagent (potassium tetraiodomercurate), one-dimensional polymer (salts of HgNH2+)n), etc.

Packaging

Plastic bottle

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

POTENTIOMETRIC-KINETIC METHOD FOR THE DETERMINATION OF MERCURY (II) BY USING ORTHO ANISIDINE
Rao D Malleswara, et al.
International journal of plant animal and environmental sciences (2012)
Mercury-impacted scrap metal: Source and nature of the mercury
Finster ME, et al.
Journal of Environmental Management, 161, 303-308 (2015)
Organomercury compounds in organic synthesis, 22 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service