Skip to Content
Merck
All Photos(2)

Key Documents

1.04933

Supelco

Potassium chloride

for analysis (<= 0.005% Br) EMSURE® ACS,ISO,Reag. Ph Eur

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-211-8
NACRES:
NA.21
Assay:
99-100.5% dry basis (silver nitrate titration)
99.5-100.5% (silver nitrate titration)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

99-100.5% dry basis (silver nitrate titration)
99.5-100.5% (silver nitrate titration)

form

solid

potency

2600 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

loss

≤1% loss on drying, 105°C

pH

5.5-8.0 (25 °C, 50 g/L in H2O)

bp

1413 °C/1013 hPa

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

bulk density

1000 kg/m3

anion traces

bromide (Br-): ≤0.005%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%passes test
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.001%

cation traces

Ba:, passes test
Ca: ≤0.001%
Fe: ≤0.0002%
Mg: ≤0.0005%
Mg: ≤0.02% (alkaline earth metals (as Ca))
N: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Analysis Note

Assay (argentometric): 99.5 - 100.5 %
Assay (argentometric; calculated on dried substance): 99.0 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water): 5.5 - 8.0
Acidity or alkalinity: passes test
Bromide (Br): ≤ 0.005 %
Chlorate and Nitrate (as NO₃): ≤ 0.003 %
Iodide (I):
≤ 0.002 %
: passes test
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): passes test
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0002 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Magnesium and alkaline-earth metals (as Ca): ≤ 0.02 %
Loss on Drying (105°C): ≤ 1.0 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service