Skip to Content
Merck
All Photos(1)

Key Documents

1.03776

Supelco

Ammonium iron(III) sulfate dodecahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Ammonium iron(III) sulfate dodecahydrate, Alum iron, Ferric ammonium sulfate, Iron alum, Iron(III) ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-382-4
NACRES:
NA.21
Assay:
99.0-102.0% (iodometric)
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (iodometric)

form

solid

impurities

≤0.005% Insoluble matter

pH

1.8 (20 °C, 100 g/L in H2O)

mp

39-41 °C

bulk density

750 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate (NO3-): ≤0.01%

cation traces

Ca: ≤0.01%
Cu: ≤0.001%
Fe2+: ≤0.001%
K: ≤0.005%
Mg: ≤0.001%
Mn: ≤0.005%
Na: ≤0.01%
Pb: ≤0.0005%
Zn: ≤0.001%

storage temp.

15-25°C

SMILES string

[Fe+3].[S](=O)(=O)([O-])O.[S](=O)(=O)([O-])[O-].N.O.O.O.O.O.O.O.O.O.O.O.O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: FX0205; FX0205-1; FX0205-20; FX0205-3

Analysis Note

Assay (iodometric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.0005 %
Nitrate (NO₃): ≤ 0.01 %
Cu (Copper): ≤ 0.001 %
Fe II (Iron II): ≤ 0.001 %
Ca (Calcium): ≤ 0.01 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.001 %
Mn (Manganese): ≤ 0.005 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %
Corresponds to ACS, ISO, Reag. Ph. Eur.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service