Skip to Content
Merck
All Photos(3)

Documents

203505

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

99.997% trace metals basis

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.997% trace metals basis

form

crystalline

impurities

≤35.0 ppm Trace Metal Analysis

mp

100 °C (dec.) (lit.)

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium iron(II)sulfate is a light green to blue crystalline powder, which dissolves readily in water, but is insoluble in ethanol and acetone. The salt melts and begins to decompose at 100 °C. It is a double salt of iron sulfate and ammonium sulfate and is made by dissolving equimolar iron(II) sulfate and ammonium sulfate in water and crystallizing the product.

Application

Ammonium iron(II) sulfate hexahydrate can be used:
  • As a precursor to synthesize iron oxide nanoparticles deposited over bismuth carbonate/ACF(fiber). This material can be used as an efficient catalyst for wastewater treatment.
  • As a negative electrolyte to develop a vanadium/iron redox flow battery (V/Fe RFB).
  • To modify methyl orange dye via metal insertion reaction. The modified dye acts as a light harvester on DSSC (Dye-Sensitized Solar Cell).
  • As a dopant to prepare nano-titania catalyst by the sol-gel method. The doping with (NH4)2Fe(SO4)2·6H2O leads to photocatalytic activation of titania in the visible light range.


Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sheng-Hua Ye et al.
Angewandte Chemie (International ed. in English), 57(10), 2672-2676 (2018-02-09)
Iron-substituted CoOOH porous nanosheet arrays grown on carbon fiber cloth (denoted as Fex Co1-x OOH PNSAs/CFC, 0≤x≤0.33) with 3D hierarchical structures are synthesized by in situ anodic oxidation of α-Co(OH)2 NSAs/CFC in solution of 0.01 m (NH4 )2 Fe(SO4 )2 . X-ray
Yu-Jung Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 520-524 (2005-04-07)
This study examined whether iron accumulated in ferritin-producing recombinant microbes is bioavailable to rats with iron deficiency. Rats induced with iron deficiency were treated with iron preparations of ferrous ammonium sulfate, horse spleen ferritin, control yeast, and ferritin-producing recombinant yeast
Kazumasa Kumamoto et al.
Free radical research, 39(9), 987-993 (2005-08-10)
Identification of free radicals was performed for the reaction mixtures of autoxidized 1,2-dilinoleoylphosphatidylcholine (DLPC) with ferrous ions (or DLPC hydroperoxide with ferrous ions) and of DLPC with soybean lipoxygenase using electron spin resonance (ESR), high performance liquid chromatography (HPLC)--ESR and
Mayara Marinovic Baccan et al.
Journal of inorganic biochemistry, 107(1), 34-39 (2011-12-17)
The antioxidant activity of flavonoids may involve their ability to complex body iron in non-redox-active forms. In this study, it was found that the catechol flavonoids rutin and quercetin are able to suppress redox-active labile plasma iron (LPI) in both
Xianwen Mi et al.
Analytical chemistry, 84(2), 939-946 (2011-12-07)
The key factors that control the spread and mortality rate of tuberculosis (TB) are rapid detection and diagnosis. However, the current detection of Mycobacterium tuberculosis (M. tuberculosis) cannot meet the recommended requirements for clinical diagnosis in turnaround time. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service