Skip to Content
Merck
All Photos(1)

Key Documents

1.03061

Supelco

Dimethylglyoxime disodium salt octahydrate

GR for analysis

Synonym(s):

Dimethylglyoxime disodium salt octahydrate, 2,3-Butanedionedioxime disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CNONa)2 · 8 H2O
CAS Number:
Molecular Weight:
304.20
UNSPSC Code:
12352107
EC Index Number:
262-523-2

Quality Level

form

solid

bulk density

300 kg/m3

storage temp.

15-25°C

InChI

1S/C4H8N2O2.2Na.8H2O/c1-3(5-7)4(2)6-8;;;;;;;;;;/h7-8H,1-2H3;;;8*1H2/q;2*+1;;;;;;;;/p-2/b5-3+,6-4+;;;;;;;;;;

InChI key

IJJDRGUPRATWQX-NPVLJBOBSA-L

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99
Identity (IR-spectrum): conforms
Substances insoluble in water: conforms
Water (according to Karl Fischer): 44 - 48
Sensitivity test (at Nickel): conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service