Skip to Content
Merck
All Photos(1)

Key Documents

40390

Sigma-Aldrich

Dimethylglyoxime

puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)

Synonym(s):

2,3-Butanedione dioxime, Diacetyldioxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=NOH)C(=NOH)CH3
CAS Number:
Molecular Weight:
116.12
Beilstein:
506731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (TLC)

form

solid

quality

for the detection of Ni

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.02% insoluble matter in ethanol

ign. residue

≤0.05% (as SO4)

mp

240-241 °C (lit.)

solubility

water: soluble 0.1 g/10 mL, clear, colorless

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

functional group

amine
oxime

SMILES string

CC(=N/O)\C(C)=N\O

InChI

1S/C4H8N2O2/c1-3(5-7)4(2)6-8/h7-8H,1-2H3/b5-3+,6-4+

InChI key

JGUQDUKBUKFFRO-GGWOSOGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethylglyoxime (DMG) is a complexing ligand. Dimethylglyoxime forms a number of mixed ligand complexes with N-acetylglycine with metals such as VO(IV), Ni(II), Zn(II), Pd(II), Cd(II) and Pb(II). It is a useful reagent for the spectrophotometric determination of Co(II), Fe(II), Ni(II), Pd(II) and Re(VII).

Application

Dimethylglyoxime (DMG) was used as complexing agent for cobalt(II) and nickel(II) metals for the analysis of brine for trace metals by adsorptive stripping voltammetry. DMG was used in a study to develop a new DMG containing modified sol-gel carbon composite electrode for voltammetric sensing of heavy metals.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of adsorptive stripping voltammetry (AdSV) for the analysis of trace metals in brine.
Romanus A, et al.
Fresenius Journal of Analytical Chemistry, 340(6), 363-370 (1991)
Preparation and Spectral Properties of Mixed-Ligand Complexes of VO (IV), Ni (II), Zn (II), Pd (II), Cd (II) and Pb (II) with Dimethylglyoxime and N-Acetylglycine.
Shaker SA
Journal of Chemistry, 7(S1), S580-S586 (2010)
Dimethylglyoxime doped sol-gel carbon composite voltammetric sensor for trace nickel.
Wang J, et al.
Electroanalysis, 9(9), 689-692 (1997)
Katherine E Heim et al.
Contact dermatitis, 60(2), 100-105 (2009-02-12)
Cutaneous nickel allergy in the very young is not well documented or characterized. A significant number of individuals are nickel sensitized by their mid-teenage years. Recent studies suggest that children may become sensitized to nickel at an early age. The
Tina Suneja et al.
Dermatitis : contact, atopic, occupational, drug, 18(4), 208-211 (2007-11-21)
Nickel is a common cause of allergic contact dermatitis and is associated with metal buttons and snaps on blue jeans. To determine the prevalence of nickel-positive metal buttons on blue jeans and the efficacy of nail polish in the prevention

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service