Skip to Content
Merck
All Photos(1)

Documents

1.02241

Sigma-Aldrich

Casein from bovine milk

soluble in alkali, Technipur®

Synonym(s):

Caseinogen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.21

Quality Level

mp

280 °C (dec.) (lit.)

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Casein is a collective term given for a family of phosphoproteins generally found in the milk of mammals. It has wide applications in the paints, medical, plastics, and food industries. It is used to form self-healing-protective coatings. Edible casein films are used in food packaging applications as a good barrier to oxygen to protect products from oxidation. The Casein-based adhesive is used as a bottle-labeling adhesive for beer and soda bottles.

Application

Casein can be used as:
  • An adhesive in the preparation of magnesium composite applicable as a fire retardant coating used in the wood industry.
  • A superplasticizer in the self-leveling underlayment (SLU) formulation.

Biochem/physiol Actions

Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milk protein polymer and its application in environmentally safe adhesives
Guo M and Wang G
Polymers, 8(9), 324-324 (2016)
Casein as a natural protein to inhibit the corrosion of mild steel in HCl solution
Rabizadeh T and Asl SK
Journal of Molecular Liquids, 276, 694-704 (2019)
Milk protein polymer and its application in environmentally safe adhesives
Guo M and Wang G
Polymer, 8(9), 324-324 (2016)
Casein films: the effects of formulation, environmental conditions and the addition of citric pectin on the structure and mechanical properties
Bonnaillie LM, et al.
Polymer, 6(7), 2018-2036 (2014)
Potential of casein as a carrier for biologically active agents
Glab TK and Boratynski J
Topics in current chemistry (Journal), 375(4), 1-20 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service