Skip to Content
Merck
All Photos(2)

Key Documents

06-933

Sigma-Aldrich

Anti-acetyl-Lysine Antibody

serum, Upstate®

Synonym(s):

Acetylated Lysine Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

acetylation (Lys)

Gene Information

human ... CECR2(27443)

General description

In the nucleus, DNA is tightly packed into nucleosomes generating an environment which is highly repressive towards DNA processes such as transcription. Acetylation of lysine residues within proteins has emerged as an important mechanism used by cells to overcome this repression. The acetylation of non-histone proteins such as transcription factors, as well as histones appears to be involved in this process. Acetylation may result in structural transitions as well as specific signaling within discrete chromatin domains. The role of acetylation in intracellular signaling has been inferred from the binding of acetylated peptides by the conserved bromodomain. Furthermore, recent findings suggest that bromodomain/acetylated-lysine recognition can serve as a regulatory mechanism in protein-protein interactions in numerous cellular processes such as chromatin remodeling and transcriptional activation.

Specificity

Acetyl-lysine containing proteins including histones, from sodium butyrate or Trichostatin A treated cells, p53 acetylated in vitro by p300 or PCAF, and chemically acetylated BSA.
Broad species reactivity expected.

Immunogen

A mixture of chemically acetylated antigens.

Application

Anti-acetyl-Lysine Antibody is a high quality Rabbit Polyclonal Antibody for the detection of acetyl-Lysine & has been validated in WB & IP.
Immunoprecipitation: 5 μL of a previous lot immunoprecipitated in vitro acetylated recombinant p300.

Western Blot Analysis: A 1:1000 dilution of the previous lot of the antibody detected:
• Acetylated histones and to a lesser degree, other proteins in RIPA lysate from trichostatin A (5 μM) treated Cos-1 cells. Trichostatin A, an inhibitor of deacetylases also, was used to increase the level of acetylated histones. (Panel B).
• GST-p53 (100 ng) acetylated in vitro with either recombinant p300 or PCAF. (Panel C).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by western blot on acid-extracted proteins from sodium butyrate treated HeLa cells, chemically acetylated BSA, acetylated histones and to a lesser degree, other proteins in trichostatin A treated Cos-1 cell lysate.

Western Blot Analysis: 1:1000-1:2000 dilution of this antibody detected:
• Acetylated histones in acid-extracted proteins from sodium butyrate (5 mM) treated HeLa cells. Sodium butyrate, an inhibitor of deacetylases, was used to increase the level of acetylated histones. (Panel A).
• Chemically acetylated BSA (10 ng) in a dot blot assay.

Target description

Dependent upon the molecular weight of the lysine acetylated protein being detected.

Physical form

Antiserum
Rabbit IgG serum containing 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
TSA-treated HeLa and NIH/3T3 cells, osteosarcoma tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibodies immobilized as arrays to profile protein post-translational modifications in mammalian cells.
Ivanov, SS; Chung, AS; Yuan, ZL; Guan, YJ; Sachs, KV; Reichner, JS; Chin, YE
Molecular and Cellular Proteomics null
Myc overexpression brings out unexpected antiapoptotic effects of miR-34a.
Sotillo, E; Laver, T; Mellert, H; Schelter, JM; Cleary, MA; McMahon, S; Thomas-Tikhonenko, A
Oncogene null
Cell cycle-dependent acetylation of Rb2/p130 in NIH3T3 cells.
Schwarze, F; Meraner, J; Lechner, M; Loidl, A; Stasyk, T; Laich, A; Loidl, P
Oncogene null
Identifying acetylated proteins in mitosis.
Carol Chuang,Li-Yuan Yu-Lee
Methods in Molecular Biology null
Multiple roles for acetylation in the interaction of p300 HAT with ATF-2.
Karanam, B; Wang, L; Wang, D; Liu, X; Marmorstein, R; Cotter, R; Cole, PA
Biochemistry null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service