Skip to Content
Merck
All Photos(1)

Key Documents

05-632

Sigma-Aldrich

Anti-14-3-3σ Antibody, clone CS112-2A8

clone CS112-2A8, Upstate®, from mouse

Synonym(s):

Stratifin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CS112-2A8, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SFN(2810)
mouse ... Sfn(55948)
rat ... Sfn(313017)

Specificity

Recognizes 14-3-3σ, does not cross-react with other 14-3-3 isoforms.

Immunogen

GST fusion protein corresponding to full-length human 14-3-3 sigma

Application

Detect 14-3-3σ using this Anti-14-3-3σ Antibody, clone CS112-2A8 validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on RIPA lysates of EGF-stimulated A431 cells

Target description

30 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Santana et al.
Journal of dairy science, 106(9), 6060-6079 (2023-07-21)
The aim of this study was to evaluate the inclusion of alfalfa grazing during 8 h continuous or partitioned in 2 separated sessions of 4 h after each milking, on nutrient intake, digestibility, ruminal fermentation, feeding behavior, milk production, milk
Claudio A Pozo et al.
Journal of dairy science, 105(6), 4987-5003 (2022-04-23)
Our aim was to evaluate whether increasing soluble carbohydrates in the herbage by changing the time of the grazing session or including Acacia mearnsii tannin in the diet would affect intake, digestion, N partitioning, and productive performance of dairy cows
D E Oliveira et al.
Journal of dairy science, 95(3), 1437-1446 (2012-03-01)
Feeding conjugated linoleic acid (CLA) in a rumen-inert form to dairy ewes has been shown to increase milk production, alter milk composition, and increase the milk fat CLA content. However, few studies have tested ruminally unprotected CLA sources. The objective
C Fernández et al.
Journal of dairy science, 104(7), 7830-7844 (2021-04-19)
The aim of this study was to evaluate the effects of incorporating rice straw and orange leaves into the diets for goats. Ten Murciano-Granadina goats at mid lactation weighing 45 ± 0.3 kg were used in a crossover design. Two
D Fernandes et al.
Animal : an international journal of animal bioscience, 8(4), 587-595 (2014-03-19)
Feeding dietary supplements containing trans-10, cis-12-conjugated linoleic acid (t10,c12-CLA) has been shown to induce milk fat depression in cows, ewes and goats. However, the magnitude of the response is apparently less pronounced in lactating goats. The objective of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service