Skip to Content
Merck
All Photos(1)

Key Documents

06-272

Sigma-Aldrich

Anti-JAK1 Antibody

serum, Upstate®

Synonym(s):

Anti-AIIDE, Anti-JAK1B, Anti-JTK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rabbit, hamster, mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JAK1(3716)

Specificity

JAK1

Immunogen

HPLC-purified (>98%) peptide corresponding to 21 amino acids of human JAK1 (KTLIEKERFYESRCRPVTPS-C)

Application

Anti-JAK1 Antibody detects level of JAK1 & has been published & validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

routinely evaluated by immunoblot on RIPA lysate from mouse CTLL lymphocytes, TF-1 erythroleukemic cells or mouse thymus

Target description

130 kDa

Physical form

Antiserum
whole rabbit antiserum containing 0.05% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C F Lai et al.
The Journal of biological chemistry, 274(12), 7793-7802 (1999-03-13)
Interleukin (IL)-6 is a major regulator of hepatic acute-phase plasma protein (APP) genes. The membrane-proximal 133-amino acid cytoplasmic domain of glycoprotein (gp) 130, containing one copy of the Box3 motif, is sufficient to transmit a productive signal to endogenous APP
Intermediate affinity interleukin-2 receptor mediates survival via a phosphatidylinositol 3-kinase-dependent pathway
Gonzalez-Garcia, A., et al
The Journal of Biological Chemistry, 272, 10220-10226 (1997)
Thrombopoietin induces tyrosine phosphorylation and activation of the Janus kinase, JAK2.
Tortolani, P J, et al.
Blood, 85, 3444-3451 (1995)
Fen Zhu et al.
Molecular cancer research : MCR, 15(3), 348-357 (2016-12-30)
JAKs are non-receptor tyrosine kinases that are generally found in association with cytokine receptors. In the canonical pathway, roles of JAKs have well been established in activating STATs in response to cytokine stimulation to modulate gene transcription. In contrast, a
Svantje Sobotta et al.
Frontiers in physiology, 8, 775-775 (2017-10-25)
IL-6 is a central mediator of the immediate induction of hepatic acute phase proteins (APP) in the liver during infection and after injury, but increased IL-6 activity has been associated with multiple pathological conditions. In hepatocytes, IL-6 activates JAK1-STAT3 signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service