Skip to Content
Merck
All Photos(1)

Key Documents

05-379

Sigma-Aldrich

Anti-E2F-1 Antibody, clones KH20 and KH95

Upstate®, from mouse

Synonym(s):

Anti-E2F-1, Anti-RBAP1, Anti-RBBP3, Anti-RBP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KH20, monoclonal
KH95, monoclonal

species reactivity

Xenopus, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... E2F1(1869)

General description

E2F-1 was identified as a cellular protein with DNA binding activity associated with the adenovirus E2 gene promoter. The original member of the E2F family of transcription factors, now consisting of five, was identified through its association with the retinoblastoma protein, which controls its activity. E2F-1 is cell cycle regulated with very low levels in early G1 then increasing levels as cells move from G1 to S with highest levels of protein at the G1/S boundary, which is consistent with its role in S-phase entry.

Specificity

Recognizes E2F-1.

Immunogen

Epitope: The epitope for KH20 has been mapped to amino acids 1-89; KH95 to amino acids 342-386.
GST fusion protein corresponding to full-length human E2F-1.

Application

Anti-E2F-1 Antibody, clones KH20 & KH95 is a high quality Mouse Monoclonal Antibody for the detection of E2F-1 & has been validated in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Immunoblot Analysis: 0.5-2μg/ml of this antibody detected E2F-1 in a HeLa nuclear extract.
Immunoprecipitation: 4μg of this antibody immunoprecipitated E2F-1 from HeLa nuclear extract.
Gel Shift: The KH20 clone has been shown to gel shift the E2F-1/DP-1/DNA complex (Helin, 1993; Wu, 2000).

Target description

55 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Xu et al.
Molecular cancer research : MCR, 14(2), 163-172 (2015-12-15)
Overexpression of EZH2 is frequently linked to the advanced and metastatic stage of cancers. The mechanisms of its oncogenic function can be context specific, and may vary depending on the protein complexes that EZH2 interacts with. To identify novel transcriptional
Expression of Dmp1 in specific differentiated, nonproliferating cells and its regulation by E2Fs.
Mallakin, A; Taneja, P; Matise, LA; Willingham, MC; Inoue, K
Oncogene null
Protein tyrosine phosphatase receptor-type O (PTPRO) is co-regulated by E2F1 and miR-17-92.
Xin Xu, Yan Hong, Chenfei Kong, Liang Xu, Jiang Tan, Qian Liang, Baiqu Huang, Jun Lu
Febs Letters null
Yibo Xue et al.
Nature communications, 10(1), 557-557 (2019-02-06)
Tumor suppressor SMARCA4 (BRG1), a key SWI/SNF chromatin remodeling gene, is frequently inactivated in cancers and is not directly druggable. We recently uncovered that SMARCA4 loss in an ovarian cancer subtype causes cyclin D1 deficiency leading to susceptibility to CDK4/6
E2F1 contributes to the transcriptional activation of the KIR3DL1 gene.
Gao, XN; Yu, L
Biochemical and biophysical research communications null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service