Skip to Content
Merck
All Photos(6)

Documents

05-1242

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4

clone 6F12-H4, from mouse

Synonym(s):

H3K9me3, Histone H3 (tri methyl K9), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6F12-H4, monoclonal

species reactivity

mouse, human

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunofluorescence: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys9)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the "histone code" hypothesis). Trimethylation of histone H3 on Lys9 (H3K9me3) is one of the most highly studied epigenetic marks. H3K9me3 functions in the repression of euchromatic genes, and in epigenetic control of heterochromatin assembly, most likely via acting as a recognition motif for the binding of chromatin-associated proteins, such as Swi6 or HP1α/β. The enzymes responsible for H3K9me3 formation are SUV39H1 and SUV39H2.

Specificity

Based on sequence homology, broad species cross-reactivity is expected with all mammals, Drosophila, Xenopus, and Arabidopsis.

Application

Chromatin Immunoprecipitation (ChIP):
Representative data from a previous lot. Sonicated 3T3 L1 chromatin was subjected to chromatin immunoprecipitation using anti- trimethyl-histone H3 (Lys9) and the Magna ChIP G (Cat. #17-611) Kit. Successful immunoprecipitation of trimethylhistone H3 (Lys9) associated DNA fragments was verified by qPCR using primers flanking the p16 promoter.

Peptide Inhibition Analysis:
Peptide blocking assay demonstrates distinct preference of the antibody for the trimethyl form vs. the dimethyl form.


Chromatin Immunoprecipitation (ChIP):
ChIP analysis of known chromosomal Suv39h targets (H3K9me3 in major satellites, mouseES cells).


Dot Blot Analysis:
Dot-blot analysis demonstrating specificity of anti-H3K9me3, clone 6F12-H4 for trimethyl Lys9 of histone H3.
Use Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4 (mouse monoclonal antibody) validated in ChIP, PIA, IF, DB, ChIP-seq, WB to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).

Quality

Routinely evaluated by Western Blot on HeLa acid extracts.

Western Blot Analysis: A 0.5 – 5 μg dilution of this lot detected trimethyl histone H3 (Lys9) in HeLa acid extracts.

Target description

~17 kDa

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Yong Yu et al.
Molecular biology reports, 39(9), 8891-8898 (2012-06-19)
Diabetic patients continue to develop inflammation and cardiovascular complication even after achieving glycemic control, suggesting a "metabolic memory". Metabolic memory is a major challenge in the treatment of diabetic complication, and the mechanisms underlying metabolic memory are not clear. Recent
Melissa Suter et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(2), 714-726 (2010-11-26)
The effect of in utero exposure to a maternal high-fat diet on the peripheral circadian system of the fetus is unknown. Using mRNA copy number analysis, we report that the components of the peripheral circadian machinery are transcribed in the
Adam E Handel et al.
Frontiers in immunology, 9, 2120-2120 (2018-10-05)
Thymic epithelial cells (TEC) effect crucial roles in thymopoiesis including the control of negative thymocyte selection. This process depends on their capacity to express promiscuously genes encoding tissue-restricted antigens. This competence is accomplished in medullary TEC (mTEC) in part by
Xu Wang et al.
The EMBO journal, 36(9), 1243-1260 (2017-03-23)
Enhancer of zeste homolog 2 (EZH2) has been characterized as a critical oncogene and a promising drug target in human malignant tumors. The current EZH2 inhibitors strongly suppress the enhanced enzymatic function of mutant EZH2 in some lymphomas. However, the
Emilie Lukášová et al.
FEBS open bio, 9(5), 870-890 (2019-04-15)
Replication stress (RS) is a major driver of genomic instability and tumorigenesis. Here, we investigated whether RS induced by the nucleotide analog fludarabine and specific kinase inhibitors [e.g. targeting checkpoint kinase 1 (Chk1) or ataxia telangiectasia and Rad3-related (ATR)] led

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service