Skip to Content
Merck
All Photos(3)

Key Documents

07-523

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys9) Antibody

serum, Upstate®

Synonym(s):

H3K9me3, Histone H3 (tri methyl K9), H3 histone, family 3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

trimethylation (Lys9)

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.

Specificity

A broad species cross-reactivity is expected.
Recognizes Histone H3 containing trimethyl-lysine 9 and, to a lesser extent, dimethyl-lysine 9. MW ~17 kDa.

Immunogen

BSA-conjugated, synthetic peptide containing the sequence …AR[me3K]S… in which me3K corresponds to trimethyl-lysine 9 of human Histone H3.
Epitope: Trimethyl Lys9

Application

Anti-trimethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9) & has been validated in DB, PIA & WB.
Peptide Inhibition Analysis:
10 μM of a Histone H3 peptide containing trimethyl-lysine 9 abolished detection of Histone H3 by anti-trimethyl Histone H3 (Lys9) in western blot analysis of HeLa acid extracts (Figure B, lane 4). Slight signal reduction was observed with 10 μM of a Histone H3 peptide containing dimethyl-lysine 9.

Peptide Dot Blot Analysis:
A 1:2000 dilution of a previous lot was incubated with serial dilutions of Histone H3 peptides containing various modifications spotted onto PVDF. Slight cross-reactivity with Histone H3 peptide containing dimethyl-lysine 9 was detected.

Beadlyte Histone Peptide Assay:
1:2000-1:6000 dilutions of a previous lot were incubated with Histone H3 peptides containing various modifications conjugated to Luminex microspheres. Slight cross-reactivity with Histone H3 peptides containing dimethyl-lysine 9 and trimethyl-lysine 27 was detected.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by western blot in acid-extracted proteins from HeLa cells.

Western Blot Analysis:
1:500-1:5000 dilution of this antibody detected methylated Histone H3 (Lys9) in acid-extracted proteins from HeLa cells. The antibody did not detect unmethylated recombinant Histone H3 (Catalog # 14-411).

Target description

~17 kDa

Physical form

Immunodepleted Serum
Rabbit polyclonal antiserum IgG in buffer containing 0.05% sodium azide before the addition of 30% glycerol.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Acid extracted proteins from HeLa cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouse homolog of SALL1, a causative gene for Townes-Brocks syndrome, binds to A/T-rich sequences in pericentric heterochromatin via its C-terminal zinc finger domains.
Kazunari Yamashita,Akira Sato,Makoto Asashima,Pi-Chao Wang,Ryuichi Nishinakamura
Genes Cells null
Ring-like distribution of constitutive heterochromatin in bovine senescent cells.
Pichugin, A; Beaujean, N; Vignon, X; Vassetzky, Y
Testing null
Shouhong Guang et al.
Nature, 465(7301), 1097-1101 (2010-06-15)
Eukaryotic cells express a wide variety of endogenous small regulatory RNAs that regulate heterochromatin formation, developmental timing, defence against parasitic nucleic acids and genome rearrangement. Many small regulatory RNAs are thought to function in nuclei. For instance, in plants and
Anne M Best et al.
Journal of biomedicine & biotechnology, 2011, 868095-868095 (2011-01-15)
Epigenetic pathways help control the expression of genes. In cancer and other diseases, aberrant silencing or overexpression of genes, such as those that control cell growth, can greatly contribute to pathogenesis. Access to these genes by the transcriptional machinery is
Cortical excitatory neurons become protected from cell division during neurogenesis in an Rb family-dependent manner.
Oshikawa, M; Okada, K; Nakajima, K; Ajioka, I
Development null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service