Skip to Content
Merck
All Photos(3)

Key Documents

04-792

Sigma-Aldrich

Anti-phospho-CENP-A (Ser7) Antibody, clone NL41, rabbit monoclonal

culture supernatant, clone NL41, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

NL41, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer7)

Gene Information

human ... CENPA(1058)

Related Categories

General description

CENP-A is a variant version of histone H3 found only at centromeres. It is phosphorylated at serine 7 during mitosis.
Centromere protein A (CENP-A) is a 17 kDa centromere-specific histone variant with 62% amino acids homology to the C-terminal of histone H3. Localized in the centromere, it plays a central role in the centromere-specific chromatin formation. The depletion of histone H3 at the CENP-A binding domain suggests CENP-A to be a possible replacement for histone H3 in the packaging process of α-satellite DNA into primary chromation structure. CENP-A is essential in the formation of specialized nucleosomes at the centromere, implicating CENP-A as a centromere-specific epigenetic marker.

Specificity

CENP-A phosphorylated on serine 7
Others not tested.

Immunogen

peptide containing the sequence RRpSRK in which pS is phospho-serine corresponding to amino acid 7 of human CENP-A (centromere protein A)

Application

Anti-phospho-CENP-A (Ser7) Antibody, clone NL41 is a high quality Rabbit Monoclonal Antibody for the detection of phospho-CENP-A (Ser7) & has been validated in Mplex, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on acid extracted proteins from colcemid-arrested HeLa cells (Catalog #17-306)

Target description

~17kDa

Linkage

Replaces: 05-792

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Acid extracted proteins from colcemid-arrested HeLa cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna P Baron et al.
eLife, 5 (2016-02-18)
The kinase Bub1 functions in the spindle assembly checkpoint (SAC) and in chromosome congression, but the role of its catalytic activity remains controversial. Here, we use two novel Bub1 inhibitors, BAY-320 and BAY-524, to demonstrate potent Bub1 kinase inhibition both
Aisling O'Connor et al.
Biology open, 5(1), 11-19 (2015-12-20)
During mitotic arrest induced by microtubule targeting drugs, the weakening of the spindle assembly checkpoint (SAC) allows cells to progress through the cell cycle without chromosome segregation occurring. PLK1 kinase plays a major role in mitosis and emerging evidence indicates
Asha Recino et al.
The Biochemical journal, 430(2), 207-213 (2010-07-16)
RASSF7, a member of the N-terminal Ras association domain family, has increased expression in various cancers and, on the basis of our previous work in Xenopus embryos, may be a regulator of mitosis. In the present study, we address, for
Anna De Antoni et al.
The Journal of cell biology, 199(2), 269-284 (2012-10-17)
By phosphorylating Thr3 of histone H3, Haspin promotes centromeric recruitment of the chromosome passenger complex (CPC) during mitosis. Aurora B kinase, a CPC subunit, sustains chromosome bi-orientation and the spindle assembly checkpoint (SAC). Here, we characterize the small molecule 5-iodotubercidin
Monika Raab et al.
Nature communications, 9(1), 1106-1106 (2018-03-20)
The spindle assembly checkpoint (SAC) acts as a molecular safeguard in ensuring faithful chromosome transmission during mitosis, which is regulated by a complex interplay between phosphatases and kinases including PLK1. Adenomatous polyposis coli (APC) germline mutations cause aneuploidy and are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service