Skip to Content
Merck
All Photos(2)

Documents

04-784

Sigma-Aldrich

Anti-IRS1 Antibody, clone AW58, rabbit monoclonal

culture supernatant, clone AW58, Upstate®

Synonym(s):

insulin receptor substrate 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AW58, monoclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IRS1(3667)

General description

Insulin Receptor substrate proteins are the effectors of both Insulin and IGF-initiated signaling. They share PH and PTB domains near their n-termini, and multiple Tyr phosphorylation motifs in their c-terminal regions. Proteins which bind to tyrosine-phosphorylated IRS-proteins include PI3 Kinase p85, GRB2, SHP2, Nck, Crk, and Fyn. IRS1 appears to be principally involved in IGF-signaling and cytoskeletal growth. IRS2 appears to be the critical mediator of Insulin signaling, as genetic ablation results in type II diabetes. IRS3 is expressed primarily in adipocytes and is an unusually potent activator of PI3 Kinase. IRS4 lacks the tyrosine residues which in the other IRS-proteins binds to SHP2.

Specificity

IRS1

Immunogen

Peptide corresponding to carboxy-terminal 14 amino acids (C-YASINFQKQPEDRQ) of rat liver IRS1

Application

Anti-IRS1 Antibody, clone AW58 detects level of IRS1 & has been published & validated for use in IP & WB.
Immunoblot Analysis: 1:1,000 to 1:4,000 dilutions of this lot detected IRS1 in RIPA lysates from 3T3/A31 mouse fibroblasts.

Immunoprecipitation: 5-10μl of this lot immunoprecipitated IRS1 from 500μg of 3T3/A31 RIPA lysate.

Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

routinely evaluated by immunoblot on in RIPA lysates from 3T3/A31 mouse fibroblasts

Target description

Mr ~145-165kDa

Linkage

Replaces: 05-784

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

1 year at -20°C from date of shipment

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Components of signaling pathways for insulin and insulin-like growth factor-I in muscle myoblasts and myotubes.
Lamphere, L and Lienhard, G E
Endocrinology, 131, 2196-2202 (1992)
Nicolas Drapeau et al.
American journal of physiology. Endocrinology and metabolism, 304(11), E1188-E1198 (2013-03-28)
Renal podocyte apoptosis is an early event of diabetic nephropathy progression. Insulin action is critical for podocyte survival. Previous studies demonstrated that Src homology-2 domain-containing phosphatase-1 (SHP-1) is elevated in renal cortex of type 1 diabetic mice; we hypothesized that
Ping Yang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(4), 5658-5672 (2020-02-27)
A contradictory role of CD36 in insulin resistance was found to be related to the nutrient state. Here, we examined that the physiological functions of CD36 in insulin signal transduction in mice fed a low-fat diet. CD36 deficiency led to
Xubo Chen et al.
Scientific reports, 7(1), 13248-13248 (2017-10-19)
Studies have reported attenuation of insulin resistance (IR) by improving phosphorylation of the insulin signalling pathway. However, the upstream molecular signalling pathway is still elusive. In this study, Western blot was used to evaluate the phosphorylation level of the insulin
A Takano et al.
Molecular and cellular biology, 21(15), 5050-5062 (2001-07-05)
A pathway sensitive to rapamycin, a selective inhibitor of mammalian target of rapamycin (mTOR), down-regulates effects of insulin such as activation of Akt (protein kinase B) via proteasomal degradation of insulin receptor substrate 1 (IRS-1). We report here that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service