Skip to Content
Merck
All Photos(1)

Key Documents

01-106

Sigma-Aldrich

FGF-2 / basic FGF Protein, Human recombinant

The FGF-2 / basic FGF Protein, Human recombinant is available in a 25 µg format.

Synonym(s):

Basic Fibroblast Growth Factor, Human FGF-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in Saccharomyces cerevisiae

form

liquid

manufacturer/tradename

Upstate®

technique(s)

cell culture | mammalian: suitable

input

sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Product Source: Recombinant DNA expressed in yeast (S. cerevisiae).

Application

The FGF-2 / basic FGF Protein, Human recombinant is available in a 25 µg format.

Quality

Routinely evaluated by Biological Activity Assay. Cultures of fetal bovine heart endothelial (FBHE) cells grown for five days in RD medium containing FBS, heparin, and FGF-2 showed more cells than cultures of FBHE cells grown in same medium without FGF-2.

Physical form

Heparin-Sepharose chromatography
Liquid in PBS, pH 7.4, containing 0.5% BSA and 0.1% CHAPS.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Chew et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(1), 227-240 (1997-01-01)
We analyzed the effects of two growth factors that regulate oligodendrocyte progenitor (O-2A) development on the expression of glutamate receptor (GluR) subunits in cortical O-2A cells. In the absence of growth factors, GluR1 was the AMPA subunit mRNA expressed at
G Keller et al.
Molecular and cellular biology, 13(1), 473-486 (1993-01-01)
We report that embryonic stem cells efficiently undergo differentiation in vitro to mesoderm and hematopoietic cells and that this in vitro system recapitulates days 6.5 to 7.5 of mouse hematopoietic development. Embryonic stem cells differentiated as embryoid bodies (EBs) develop
In vitro assays for neural stem cell differentiation.
Marcel M Daadi
Methods in molecular biology (Clifton, N.J.), 198, 149-155 (2002-04-16)
J C Fox et al.
The Journal of biological chemistry, 271(21), 12578-12584 (1996-05-24)
Basic fibroblast growth factor (bFGF), a potent mitogen for many cell types, is expressed by vascular smooth muscle cells and plays a prominent role in the proliferative response to vascular injury. Basic FGF has also been implicated as a survival
E Meuillet et al.
Glia, 17(3), 206-216 (1996-07-01)
Polypeptide growth factors and membrane-bound gangliosides are involved in cell signaling, including that observed in cells of neural origin. To analyze possible interactions between these two systems, we investigated the modulation of short- and long-term responses to basic fibroblast and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service