Skip to Content
Merck
All Photos(1)

Key Documents

V-006

Supelco

Valproic acid solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CCCC(CCC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NIJJYAXOARWZEE-UHFFFAOYSA-N

Gene Information

human ... ALDH5A1(7915)

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, urine drug testing, prescription monitoring or pharmaceutical research. Valproic acid, sold under trade names including Depakote®, Valparin, or Stavzor®, is an antiepileptic drug and mood stabilizer. The drug is prescribed for treatment of epilepsy, bipolar disorder, migraine headaches, and depression.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Depakote is a registered trademark of Sanofi S.A.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Stavzor is a registered trademark of Noven Therapeutics, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew D Sztajnkrycer
Journal of toxicology. Clinical toxicology, 40(6), 789-801 (2002-12-12)
Acute valproic acid intoxication is an increasing problem, accounting for more than 5000 calls to the American Association of Poison Control Centers in 2000. The purpose of this paper is to review the pharmacology and toxicology of valproic acid toxicity.
P C Ho et al.
The pharmacogenomics journal, 3(6), 335-342 (2003-11-05)
The present study investigated the effect of cytochrome P450 2C9 (CYP2C9) genetic polymorphism on the biotransformation of valproic acid (VPA) to its hepatotoxic metabolite, 4-ene-VPA, and compared that to the formation of the inactive 4-OH-VPA and 5-OH-VPA. cDNA-expressed CYP2C9(*)2 and
Wolfgang Löscher
CNS drugs, 16(10), 669-694 (2002-09-25)
Since its first marketing as an antiepileptic drug (AED) 35 years ago in France, valproate has become established worldwide as one of the most widely used AEDs in the treatment of both generalised and partial seizures in adults and children.
Emily C Bell et al.
Human psychopharmacology, 20(6), 415-424 (2005-08-18)
Previous functional imaging studies have shown altered brain activity during cognitive task performance in bipolar patients. However, the fact that these patients are often on medication makes it unclear to what extent these changes reflect treatment effects. This study aims
Chi-Tso Chiu et al.
Pharmacological reviews, 65(1), 105-142 (2013-01-10)
The mood stabilizers lithium and valproic acid (VPA) are traditionally used to treat bipolar disorder (BD), a severe mental illness arising from complex interactions between genes and environment that drive deficits in cellular plasticity and resiliency. The therapeutic potential of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service