Skip to Content
Merck
All Photos(1)

Key Documents

A-064

Supelco

Acetaminophen solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)Nc1ccc(O)cc1

InChI

1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)

InChI key

RZVAJINKPMORJF-UHFFFAOYSA-N

Gene Information

General description

An analytical reference standard suitable for use as starting material for calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to P450 studies and other pharmaceutical research. Acetaminophen, or Paracetamol, is a common over-the-counter COX inhibitor drug used for pain relief and fever reduction.


This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application



  • Enhanced removal of acetaminophen from aqueous solutions: A study utilized kumquat peel-derived biochar supporting zeolitic imidazole framework-67 (ZIF-67) to improve peracetic acid activation for acetaminophen removal, demonstrating an innovative approach to wastewater treatment (Nguyen et al., 2024).


  • Co-doped biochar for acetaminophen degradation: Research on Cu/N co-doped biochar activating peroxymonosulfate (PMS) revealed a non-radical degradation pathway for acetaminophen dominated by singlet oxygen and electron transfer, providing insights into sustainable environmental cleanup technologies (Wu et al., 2024).


  • Biodegradation of pharmaceuticals including acetaminophen: A study on anaerobic co-metabolic biodegradation of pharmaceuticals and personal care products highlighted glycerol fermentation as a driving mechanism, offering a feasible strategy for managing environmental contamination from substances like acetaminophen (Carneiro et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Munsterhjelm et al.
Anesthesiology, 103(4), 712-717 (2005-09-30)
Acetaminophen (paracetamol) is widely used for postoperative analgesia. Its mechanism of action is inhibition of prostaglandin synthesis in the central nervous system, and acetaminophen is traditionally not considered to influence platelet function. The authors studied the dose-dependent inhibition of platelet
Garry G Graham et al.
Inflammopharmacology, 21(3), 201-232 (2013-05-31)
Paracetamol is used worldwide for its analgesic and antipyretic actions. It has a spectrum of action similar to that of NSAIDs and resembles particularly the COX-2 selective inhibitors. Paracetamol is, on average, a weaker analgesic than NSAIDs or COX-2 selective
S M de Morais et al.
Gastroenterology, 102(2), 577-586 (1992-02-01)
Gilbert's syndrome occurs in 5%-7% of the human population and is caused by an inherited deficiency in the glucuronidation of endogenous bilirubin, resulting in its accumulation and jaundice. The authors of the present study have previously shown that rats with
Anne M C M Doedée et al.
PloS one, 9(6), e98175-e98175 (2014-06-05)
Worldwide, paracetamol is administered as a remedy for complaints that occur after vaccination. Recently published results indicate that paracetamol inhibits the vaccination response in infants when given prior to vaccination. The goal of this study was to establish whether paracetamol
Karel Allegaert et al.
Archives of disease in childhood, 98(6), 462-466 (2013-04-23)
There remains a need for alternative medical treatments for patent ductus arteriosus (PDA) closure in extreme preterm neonates because of therapeutic failure and adverse effects associated with non-selective cyclo-oxygenase inhibitors. Reports of an association between paracetamol exposure and PDA closure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service