Skip to Content
Merck
All Photos(1)

Key Documents

P-080

Supelco

Phenazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2OBrCl
CAS Number:
Molecular Weight:
349.61
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Clc1ccccc1C2=NCC(=O)Nc3ccc(Br)cc23

InChI

1S/C15H10BrClN2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20)

InChI key

CGMJQQJSWIRRRL-UHFFFAOYSA-N

General description

Phenazepam, a benzodiazepine drug produced in Russia, is used in Europe as an anxiolytic, anticonvulsant, and muscle relaxant. Although not used therapeutically in the US, the drug can be purchased on the internet for “laboratory research” purposes. Cases of abuse have been reported from European countries such as Sweden and Russia to states including New York, Kentucky, North Carolina, and Georgia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G G Neznamov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(5), 6-12 (2011-08-04)
Differences between phenazepam formulations were found in the clinical study of drug effects on the psychophysiological state in patients with anxiety disorder. Phenazepam did not cause negative changes of psychophysiological parameters. In patients with anxiety disorders, the most significant positive
L I Larentsova et al.
Bulletin of experimental biology and medicine, 148(2), 340-342 (2009-12-23)
The use of tenoten and tenoten (pediatric formulation) for premedication in adults and children before dental treatment reduces patient anxiety; the effect is comparable with that of classical anxiolytics and is not accompanied by side effects. These preparations can be
N G Bogdan et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(8), 8-12 (2012-01-12)
The influence of the selective anxiolytic drug afobazole in a single dose of 10 mg on the psychophysiological functions of healthy volunteers was studied in laboratory experiments. It is established that afobazole optimizes some psychophysiological characteristics of stress-labile individuals and
L N Pavlov et al.
Radiatsionnaia biologiia, radioecologiia, 52(4), 388-393 (2012-10-05)
Effects of EMR on the behavior of Wistar rats (196 males, 180-240 g of mass) under the conflict of opposed motivations: strong positive, drinking, motivation, and strong negative, pain, motivation were studied. The animals were exposed to low intensity EMR
[Pharmacologic correction of psycho-emotional status of patients with thyroid nodules in the preoperative period].
E M Zvukova et al.
Voprosy onkologii, 56(6), 722-725 (2010-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service