Skip to Content
Merck
All Photos(1)

Key Documents

L-019

Supelco

Lamotrigine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Nc1nnc(c(N)n1)-c2cccc(Cl)c2Cl

InChI

1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)

InChI key

PYZRQGJRPPTADH-UHFFFAOYSA-N

General description

Lamotrigine is an anticonvulsant approved for treatment of epilepsy and bipolar disorder. Sold under the trade name Lamictal®, this phenyltriazine analog also is used off-label for treatment of clinical depression. This Snap-N-Spike® Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Lamictal is a registered trademark of GlaxoSmithKline LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shai Sandalon et al.
Experimental eye research, 115, 47-56 (2013-07-03)
Voltage gated sodium channels (Nav), are proposed mediators of neuronal damage in ischemic and excitotoxicity disease models. We evaluated the neuroprotective effects of lamotrigine, a Nav blocker, in the acute and chronic rat ocular hypertension models. Additionally, expression of the
Stacey Shim et al.
The Journal of pharmacology and experimental therapeutics, 347(2), 487-496 (2013-08-30)
Carisbamate and lamotrigine are anticonvulsants that act on neuronal voltage-gated sodium channels. Carisbamate has shown antidepressant-like effects in animal models of depression, and lamotrigine is a mood stabilizer with a therapeutic effect in depressive episodes of patients with bipolar disorder.
Maxine Dibué et al.
Epilepsia, 54(9), 1542-1550 (2013-06-19)
Lamotrigine (LTG) is a popular modern antiepileptic drug (AED); however, its mechanism of action has yet to be fully understood, as it is known to modulate many members of several ion channel families. In heterologous systems, LTG inhibits Cav 2.3
Allyson K Friedman et al.
Science (New York, N.Y.), 344(6181), 313-319 (2014-04-20)
Typical therapies try to reverse pathogenic mechanisms. Here, we describe treatment effects achieved by enhancing depression-causing mechanisms in ventral tegmental area (VTA) dopamine (DA) neurons. In a social defeat stress model of depression, depressed (susceptible) mice display hyperactivity of VTA
Philip J Wiffen et al.
The Cochrane database of systematic reviews, 12(12), CD006044-CD006044 (2013-12-04)
This is an update of the original Cochrane review entitled Lamotrigine for acute and chronic pain published in Issue 2, 2007, and updated in Issue 2, 2011. Some antiepileptic medicines have a place in the treatment of neuropathic pain (pain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service