Skip to Content
Merck
All Photos(2)

Documents

61347

Supelco

10,11-Dihydro-10-hydroxycarbamazepine

analytical standard

Synonym(s):

10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, Licarbazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
Beilstein:
1540211
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)

InChI key

BMPDWHIDQYTSHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10,11-Dihydro-10-hydroxycarbamazepine is a 10-hydroxy derivative and an active metabolite of oxcarbazepine.
10,11-Dihydro-10-hydroxycarbamazepine is an active metabolite of the antiepileptic drug, oxcarbazepine.

Application

10,11-Dihydro-10-hydroxycarbamazepine may be used as a reference standard in the determination of 10,11-dihydro-10-hydroxycarbamazepine in biological samples using liquid chromatography, liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) and high performance liquid chromatography coupled with ultraviolet detector (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Pohl et al.
Environmental science & technology, 54(5), 2913-2921 (2020-01-29)
Carbamazepine (CBZ) is an anticonvulsant medication with highly persistent properties in the aquatic environment, where it has the potential to affect nontarget biota. Because CBZ and many other pharmaceuticals are not readily removed in conventional sewage treatment plants (STP), additional
Liquid chromatographic analysis of oxcarbazepine and its metabolites in plasma and saliva after a novel microextraction by packed sorbent procedure.
Saracino M A, et al.
Analytica Chimica Acta, 661(2), 222-228 (2010)
Simultaneous quantitative analysis of oxcarbazepine and 10, 11-dihydro-10-hydroxycarbamazepine in human plasma by liquid chromatography-electrospray tandem mass spectrometry.
Maia SdBM, et al.
Journal of Pharmaceutical and Biomedical Analysis, 45(2), 304-311 (2007)
Anne-Catherine Servais et al.
Journal of chromatography. A, 1467, 306-311 (2016-07-22)
A LC method using a chiral stationary phase (CSP) with cellulose tris(3-chloro-4-methylphenylcarbamate) as chiral selector in polar organic mode (POM) was developed for the separation of the biopharmaceutic classification system (BCS) class II chiral prodrug eslicarbazepine acetate (ESL) and its
Shengyang Zhou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1141, 122036-122036 (2020-02-29)
In this study, haptens were designed to produce highly sensitive and specific monoclonal antibodies (mAb) against carbamazepine (CBZ) and its metabolite carbamazepine-10, 11-epoxide (CBZ-EP). According to the results of our competitive enzyme-linked immunosorbent assay (ic-ELISA), the half-maximum inhibitory concentration values

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service