Skip to Content
Merck
All Photos(1)

Key Documents

C-108

Supelco

Cyclosporin D solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

1-(3-Chlorophenyl)piperazine-D8 hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

C\C=C\C[C@@H](C)[C@@H](O)[C@@H]1N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C(=O)[C@@H](NC1=O)C(C)C)C(C)C

InChI

1S/C63H113N11O12/c1-26-27-28-41(16)53(76)52-57(80)67-49(38(10)11)61(84)68(19)33-48(75)69(20)44(29-34(2)3)56(79)66-50(39(12)13)62(85)70(21)45(30-35(4)5)55(78)64-42(17)54(77)65-43(18)58(81)71(22)46(31-36(6)7)59(82)72(23)47(32-37(8)9)60(83)73(24)51(40(14)15)63(86)74(52)25/h26-27,34-47,49-53,76H,28-33H2,1-25H3,(H,64,78)(H,65,77)(H,66,79)(H,67,80)/b27-26+/t41-,42+,43-,44+,45+,46?,47+,49+,50+,51+,52+,53-/m1/s1

InChI key

ZNVBEWJRWHNZMK-UPMQNEEPSA-N

General description

An internal standard for clinical testing of Cyclosporin A by LC-MS/MS. This Certified Spiking Solution® is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within Cyclosporin A′s narrow therapeutic range. The Snap-N-Spike® format allows laboratories to eliminate weighing operations of these hazardous substances, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.

Application


  • High-Performance Liquid Chromatography for Cyclosporin D: A study developed a reversed-phase high-performance liquid chromatography method for the determination of cyclosporin A in human and mouse plasma, using Cyclosporin D as internal standard, highlighting its applications in drug discovery and bioavailability studies (Bardelmeijer et al., 2001).

  • Mass Spectrometric Analysis of Cyclosporins: Research focused on the thermospray high-performance liquid chromatography/mass spectrometric determination of cyclosporins, including Cyclosporin D, contributing to advancements in the analysis of immunosuppressant drugs and their pharmacological profiling (Abián et al., 1992).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wenzel-Seifert et al.
Journal of immunology (Baltimore, Md. : 1950), 147(6), 1940-1946 (1991-09-15)
Cyclosporin (Cs)A but not CsH inhibits activation of human lymphocytes. We studied the effects of CsA, CsD, and CsH on human neutrophil activation induced by chemoattractants and by various substances that circumvent receptor stimulation. CsH inhibited superoxide (O2-) formation induced
Synergistic effects of cyclosporin analogs--A, D, G, IMM-125--with rapamycin and/or brequinar.
B D Kahan et al.
Transplantation proceedings, 26(5), 3021-3024 (1994-10-01)
K Tanaka et al.
Pharmaceutical research, 13(7), 1073-1077 (1996-07-01)
P-glycoprotein-mediated transcellular transport of anticancer agents and the inhibitory effect of cyclosporin analogs and FK506 were investigated. The transcellular transport of daunorubicin and vinblastine by monolayers of LLC-GA5-COL150 cells which overexpressed P-glycoprotein was measured in the presence and absence of
G M Randleff-Rasmussen et al.
Transplant international : official journal of the European Society for Organ Transplantation, 2(4), 181-186 (1989-12-01)
Various methods of determining cyclosporin (CyA) levels in patients after kidney transplantation were compared. These included polyclonal antibody (pcAb-), specific and nonspecific monoclonal antibody (S- and NmcAb-) radioimmunoassays (RIA), and high performance liquid chromatography (HPLC). The results obtained by the
N W Wilkinson et al.
International journal of surgical investigation, 2(6), 491-498 (2003-04-08)
The adenoviral vector demonstrates efficient gene transfer and high transient gene expression making it an attractive vehicle for human gene therapy trials. Unfortunately, the virus stimulates a potent inflammatory immune response that limits transgene expression and makes repeat viral dosing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service