Skip to Content
Merck
All Photos(1)

Key Documents

B-006

Supelco

Butalbital solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

5-Allyl-5-isobutylbarbituric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O3
CAS Number:
Molecular Weight:
224.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CC(C)CC1(CC=C)C(=O)NC(=O)NC1=O

InChI

1S/C11H16N2O3/c1-4-5-11(6-7(2)3)8(14)12-10(16)13-9(11)15/h4,7H,1,5-6H2,2-3H3,(H2,12,13,14,15,16)

InChI key

UZVHFVZFNXBMQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butalbital is a barbiturate often combined with other medications such as acetaminophen or aspirin for the treatment of pain or codeine for headaches. Trade names of several butalbital combinations include Axocet®, Bucet, and Bupap® with acetaminophen and Axotal with aspirin. This certified reference solution is suitable for GC/MS or LC/MS applications in forensic analysis, clinical toxicology or urine drug testing.

Legal Information

Axocet is a registered trademark of Fougera Pharmaceuticals Inc.
Bupap is a registered trademark of ECR Pharmaceuticals Co., Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William B Young et al.
Current pain and headache reports, 6(2), 151-155 (2002-03-02)
In the United States analgesic-overuse headache is often caused by butalbital-containing analgesics. These agents can cause physical and psychological dependency, and dangerous withdrawal syndromes. Butalbital-containing analgesics have already been banned in several European countries. They are proven effective in tension-type
W T Chang et al.
Journal of analytical toxicology, 25(8), 659-669 (2002-01-05)
As a part of our study on the use of isotopic analogues as the internal standard (IS) for the quantitation of drug analytes, this article reports on the performance characteristics of 2H5-butalbital and 13C4-butalbital with particular focus on (1) determining
Richard G Wenzel et al.
Pharmacotherapy, 22(8), 1029-1035 (2002-08-14)
To evaluate the role of butalbital-containing products in the management of migraine. Qualitative systematic search using MEDLINE (January 1966-November 2001), review of the United States Headache Consortium's evidence-based guidelines for migraine treatment, and review of other pertinent literature. Over 28
Seymour Solomon
Current pain and headache reports, 6(2), 147-150 (2002-03-02)
Butalbital compounds are of proven efficacy in the treatment of tension headache. Decades of experience have established their value in the treatment of other mild-to-moderate headaches. Untold numbers of people rely on these medications as their drug of choice or
Karla A Moore et al.
Forensic science international, 134(2-3), 120-122 (2003-07-10)
Zaleplon ("Sonata") is a pyrazolopyrimidine derivative approved for use in the United States for the treatment of insomnia. To date, there has been little data in the toxicological literature where zaleplon has been implicated as causing a fatal intoxication, either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service