Skip to Content
Merck
All Photos(1)

Key Documents

A-053

Supelco

6-Acetylcodeine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO4
CAS Number:
Molecular Weight:
341.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccc2C[C@@H]3[C@@H]4C=C[C@H](OC(C)=O)[C@@H]5Oc1c2[C@]45CCN3C

InChI

1S/C20H23NO4/c1-11(22)24-16-7-5-13-14-10-12-4-6-15(23-3)18-17(12)20(13,19(16)25-18)8-9-21(14)2/h4-7,13-14,16,19H,8-10H2,1-3H3/t13-,14+,16-,19-,20-/m0/s1

InChI key

MFXFQKMUCYHPFQ-BKRJIHRRSA-N

General description

A Certified Snap-N-Spike® Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. 6-Acetylcodeine is an analog of codeine and is sometimes present as an impurity in street heroin.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daming Zhang et al.
Journal of forensic sciences, 49(1), 81-86 (2004-02-26)
A novel method based on GC/MS and GC for component analyses of seized illicit heroin was established by using SKF525A as an internal standard. The main components in illicit heroin products such as heroin, O3-acetylmorphine, monoacetylcodeine, and O6-acetylmorphine were determined
S G Phillips et al.
Journal of analytical toxicology, 30(6), 370-374 (2006-07-29)
A method was developed using liquid chromatography linked to atmospheric pressure ionization-tandem mass spectrometry (LC-MS-MS) for the measurement of the opiates, morphine, codeine, 6-monoacetylmorphine (6-MAM), acetylcodeine (AC), and heroin in oral fluid collected from patients attending a substance abuse clinic.
P Kintz et al.
Journal of analytical toxicology, 22(6), 425-429 (1998-10-27)
In addition to acetylmorphine (6-AM), acetylcodeine (AC) has been suggested as a marker for the use of illicit heroin. Because no procedure was available for AC testing in hair, a new method was developed for the simultaneous identification and quantitation
C L O'Neal et al.
Journal of analytical toxicology, 21(6), 427-432 (1997-11-05)
In addition to 6-monoacetylmorphine (6-MAM), acetylcodeine (AC) has been suggested as a marker for the use of illicit heroin. We report a sensitive opiate gas chromatographic-mass spectrometric assay that detects AC, diacetylmorphine, and the propionylated derivatives of codeine, morphine, 6-MAM
Frank Musshoff et al.
International journal of legal medicine, 123(5), 363-369 (2008-07-29)
One month before (T-1) and 12 months after (T12) controlled i.v. administration of pharmaceutical heroin-HCl (10-100 mg/day) in the context of a heroin maintenance program (HMP), concentrations of opiates and cocaine as well as its metabolites were determined in head

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service