Skip to Content
Merck
All Photos(1)

Key Documents

880124P

Avanti

DSPE-PEG(2000) Folate

Avanti Research - A Croda Brand 880124P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (880124P-1mg)
pkg of 1 × 10 mg (880124P-10mg)
pkg of 1 × 5 mg (880124P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880124P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CC[C@H](NC(C1=CC=C(NCC(C=N2)=NC3=C2N=C(N)NC3=O)C=C1)=O)C([O-])=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C

Application

DSPE-PEG(2000) Folate has been mixed in chloroform for folate-targeted PEGylated liposomes preparation.

Biochem/physiol Actions

Folate offers protection against megaloblastic anaemia during pregnancy.

Packaging

5 mL Amber Glass Screw Cap Vial (880124P-10mg)
5 mL Amber Glass Screw Cap Vial (880124P-1mg)
5 mL Amber Glass Screw Cap Vial (880124P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folate and human reproduction.
Tamura T and Picciano MF
American Journal of Clinical Nutrition, 83(5), 993-1016 (2006)
Microfluidic synthesis of PEG-and folate-conjugated liposomes for one-step formation of targeted stealth nanocarriers
Hood RR, et al.
Pharmaceutical Research, 30(6), 1597-1607 (2013)
A Gabizon et al.
Bioconjugate chemistry, 10(2), 289-298 (1999-03-17)
Conjugates of three components, folic acid-poly(ethylene glycol)-distearoylphosphatidylethanolamine (FA-PEG-DSPE), derived from PEG with molecular masses of 2000 and 3350 Da were synthesized by a carbodiimide-mediated coupling of FA to H2N-PEG-DSPE. The conjugates were characterized by 1H NMR, MALDI-TOF, and HPLC analysis
Yonghua Zhang et al.
Analytical biochemistry, 332(1), 168-177 (2004-08-11)
Cell membrane-associated folate receptors are selectively overexpressed in certain human tumors. The high affinity of folic acid for folate receptors provides a unique opportunity to use folic acid as a targeting ligand to deliver chemotherapeutic agents to cancer cells. Folate-tethered
Jun Wu et al.
International journal of pharmaceutics, 316(1-2), 148-153 (2006-03-28)
A novel liposomal formulation of paclitaxel targeting the folate receptor (FR) was synthesized and characterized. This formulation was designed to overcome vehicle toxicity associated with the traditional Cremophor EL-based formulation and to provide the added advantages of prolonged systemic circulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service