Skip to Content
Merck
All Photos(1)

Key Documents

860593P

Avanti

24:1 SM

Avanti Research - A Croda Brand, powder

Synonym(s):

N-nervonoyl-D-erythro-sphingosylphosphorylcholine, SM(d18:1/24:1(15Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H93N2O6P
CAS Number:
Molecular Weight:
813.22
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (860593P-25mg)
pkg of 1 × 5 mg (860593P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C47H93N2O6P/c1-6-8-10-12-14-16-18-20-21-22-23-24-25-26-27-29-31-33-35-37-39-41-47(51)48-45(44-55-56(52,53)54-43-42-49(3,4)5)46(50)40-38-36-34-32-30-28-19-17-15-13-11-9-7-2/h20-21,38,40,45-46,50H,6-19,22-37,39,41-44H2,1-5H3,(H-,48,51,52,53)/b21-20-,40-3

InChI key

WKZHECFHXLTOLJ-QYKFWSDSSA-N

Application

24:1 SM has been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions. It has also been used as a standard for the absolute quantification of epidermal lipids using liquid chromatography-tandem mass spectrometry (LC/MS/MS).

Packaging

5 mL Amber Glass Screw Cap Vial (860593P-25mg)
5 mL Amber Glass Screw Cap Vial (860593P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianing Mi et al.
Journal of agricultural and food chemistry, 66(34), 8991-8998 (2018-07-31)
Deep profiling of glycosphingolipids and sphingomyelins in wild Cordyceps was carried out by using offline chromatographic enrichment followed by ultrahigh performance liquid chromatography-ultrahigh definition-quadrupole time-of-flight mass spectrometry (UHPLC-UHD-Q-TOF-MS). A total of 119 glycosphingolipids (72 new ones) and 87 sphingomyelins (43
Srinath Govindarajan et al.
EMBO reports, 21(6), e48927-e48927 (2020-05-05)
CD1d-restricted invariant natural killer T (iNKT) cells constitute a common glycolipid-reactive innate-like T-cell subset with a broad impact on innate and adaptive immunity. While several microbial glycolipids are known to activate iNKT cells, the cellular mechanisms leading to endogenous CD1d-dependent
Zhixing Wang et al.
The Journal of investigative dermatology, 133(3), 668-676 (2012-10-26)
The stratum corneum is composed of protein-enriched corneocytes embedded in an intercellular matrix of nonpolar lipids organized as lamellar layers and giving rise to epidermal permeability barrier (EPB). EPB defects have an important role in the pathophysiology of skin diseases
Delphine Milhas et al.
FEBS letters, 584(9), 1887-1894 (2009-10-28)
The plasma membrane (PM) is a major resource for production of bioactive lipids and contains a large proportion of the cellular sphingomyelin (SM) content. Consequently, the regulation of SM levels at the PM by enzymes such as sphingomyelinase (SMase) and
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service