Skip to Content
Merck
All Photos(3)

Key Documents

857235P

Avanti

(2S)-OMPT

Avanti Research - A Croda Brand

Synonym(s):

(2S)-1-(9Z-octadecenoyl)-2-O-methyl-glycero-3-phosphothionate (ammonium salt), 1-oleoyl-2-methyl-sn-glycero-3-phosphothionate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46NO6PS
CAS Number:
Molecular Weight:
483.64
UNSPSC Code:
51191904

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (857235P-25MG)
pkg of 1 × 5 mg (857235P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH4+].CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](COP(O)([O-])=S)OC

Application

lipids research

Packaging

5 mL Amber Glass Screw Cap Vial (857235P-5MG);5 mL Amber Glass Screw Cap Vial (857235P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of LPAR3, PKC and EGFR in LPA-induced cell migration in oral squamous carcinoma cells
Brusevold IJ, et al.
BMC Cancer, 14(1), 432-432 (2014)
Pericytes prevent regression of endothelial cell tubes by accelerating metabolism of lysophosphatidic acid
Motiejunaite R, et al.
Microvascular Research, 93(2), 62-71 (2014)
Enantioselective responses to a phosphorothioate analogue of lysophosphatidic acid with LPA3 receptor-selective agonist activity.
Qian L, et al.
Journal of Medicinal Chemistry, 46, 5575-5578 (2003)
Yutaka Hasegawa et al.
The Journal of biological chemistry, 278(14), 11962-11969 (2003-01-30)
Lysophosphatidic acid (LPA) is a bioactive lysophospholipid mediator that acts through G protein-coupled receptors. Most cell lines in culture express one or more LPA receptors, making it difficult to assign a response to specific LPA receptors. Dissection of the signaling
Lian Qian et al.
Journal of medicinal chemistry, 46(26), 5575-5578 (2003-12-12)
The metabolically stabilized LPA analogue, 1-oleoyl-2-O-methyl-rac-glycerophosphothioate (OMPT), is a potent agonist for the LPA(3) G-protein-coupled receptor. A new enantiospecific synthesis of both (2R)-OMPT and (2S)-OMPT is described. Calcium release assays in both LPA(3)-transfected insect Sf9 and rat hepatoma Rh7777 cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service