Skip to Content
Merck
All Photos(2)

Key Documents

850345C

Avanti

14:0 PC (DMPC)

1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phosphocholine; DMPC; PC(14:0/14:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H72NO8P
CAS Number:
Molecular Weight:
677.93
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850345C-25mg)
pkg of 1 × 20 mL (850345C-500mg)
pkg of 2 × 20 mL (850345C-1g)
pkg of 2 × 4 mL (850345C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850345C

concentration

10 mg/mL (850345C-25mg)
25 mg/mL (850345C-1g)
25 mg/mL (850345C-200mg)
25 mg/mL (850345C-500mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@@H](OC(=O)CCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C36H72NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m0/s1

InChI key

CITHEXJVPOWHKC-UMSFTDKQSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[1] It is also an important component of the mucosal layer of the colon.[2]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

14:0 PC (DMPC) has been used in liposome preparation.[3] It is also suitable for use:
  • in the preparation of lipid bicelles for nuclear magnetic resonance (NMR) experiments[4]
  • in labeled vesicle preparation for fluorescence polarization measurements[5]
  • as internal standard (IS) for the quantitation of phosphatidylcholine (PC) by liquid chromatography with tandem mass spectrometry (LC-MS-MS)[6]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[7]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850345C-1g)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850345C-500mg)
5 mL Clear Glass Sealed Ampule (850345C-200mg)
5 mL Clear Glass Sealed Ampule (850345C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
In vitro Membrane Interaction and Liposome Fusion Assays Using Recombinant Hepatitis C Virus Envelope Protein E2
Yelamos B, et al.
Biochimica et biophysica acta. General subjects (2018)
Lipid binding by disordered proteins
Perez Y, et al.
Protocol Exchange, 10 (2013)
Integrative Medicine - E-Book (2017)
The acceleration of implant osseointegration by liposomal Wnt3a
Popelut A, et al.
Biomaterials, 31(35), 9173-9181 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service