Skip to Content
Merck
All Photos(1)

Key Documents

P54280

Sigma-Aldrich

Dipropyl sulfide

97%

Synonym(s):

Propyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2S
CAS Number:
Molecular Weight:
118.24
Beilstein:
1719002
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4487 (lit.)

bp

142-143 °C (lit.)

mp

−103 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

CCCSCCC

InChI

1S/C6H14S/c1-3-5-7-6-4-2/h3-6H2,1-2H3

InChI key

ZERULLAPCVRMCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagalingam Rajamanickam et al.
Nanotechnology, 27(23), 235404-235404 (2016-04-30)
Degradation of metal-organic halide perovskites when exposed to ambient conditions is a crucial issue that needs to be addressed for commercial viability of perovskite solar cells (PSCs). Here, a concept of encapsulating CH3NH3PbI3 perovskite crystals with a multi-functional graphene-polyaniline (PANI)
R M Nickson et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(2), 157-168 (1994-02-01)
1. Dipropyl [35S]-sulphide and dipropyl [35S]-sulphoxide were administered by gavage (4.24 mM/4 ml/kg body wt) to adult male Wistar rats following an overnight fast. 2. Urine was the major route of excretion for both compounds, with more radioactivity appearing during
X Hu et al.
Archives of biochemistry and biophysics, 340(2), 279-286 (1997-04-15)
Glutathione S-transferases (GSTs) of female A/J mouse lung have been purified and characterized for their (a) structural interrelationships, (b) substrate specificities toward the ultimate carcinogenic metabolite of benzo(a)pyrene (BP), (+)-anti-7 beta, 8 alpha-dihydroxy-9 alpha, 10 alpha-oxy-7,8,9,10-tetrahydrobenzo(a)pyrene [(+)-anti-BPDE], and (c) induction
Erna Schuhfried et al.
PloS one, 8(6), e66925-e66925 (2013-07-11)
We report on a new approach for studying fragmentation channels in Proton Transfer Reaction-Mass Spectrometry (PTR-MS), which we name primary ion depletion kinetics (PIDK). PTR-MS is a chemical ionization mass spectrometric (CIMS) technique deploying hydronium ions for the chemical ionization.
C F Williams et al.
Molecular and biochemical parasitology, 206(1-2), 20-28 (2016-03-13)
Spironucleus vortens is a protozoan parasite associated with significant mortalities in the freshwater angelfish, Pterophyllum scalare. Control of this parasite is especially problematic due to restrictions on the use of the drug of choice, metronidazole (MTZ), on fish farms. Use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service