Skip to Content
Merck
All Photos(2)

Key Documents

850330P

Avanti

11:0 PC

1,2-diundecanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(11:0/11:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60NO8P
CAS Number:
Molecular Weight:
593.77
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850330P-25mg)
pkg of 1 × 500 mg (850330P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850330P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCC)=O)COC(CCCCCCCCCC)=O)=O

InChI

1S/C30H60NO8P/c1-6-8-10-12-14-16-18-20-22-29(32)36-26-28(27-38-40(34,35)37-25-24-31(3,4)5)39-30(33)23-21-19-17-15-13-11-9-7-2/h28H,6-27H2,1-5H3

InChI key

XCCDCYFQYUARAY-UHFFFAOYSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes. It is also an important component of the mucosal layer of the colon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

11:0 PC has been used as a direct supplementation in bacteria, that are seeded onto the plates to study its effects on lipid accumulation.

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

30 mL Amber Wide Mouth Bottle with Screw Cap (850330P-500mg)
5 mL Clear Glass Sealed Ampule (850330P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Microbial metabolites regulate host lipid metabolism through NR5A-Hedgehog signalling
Lin CCJ and Wang MC
Nature Cell Biology, 19(5), 550-550 (2017)
Shi-Yuan Wen et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 69, 152690-152690 (2018-11-06)
Rhodioloside is a glucoside of tyrosol isolated from Rhodiola rosea. However, its regulating effect on hepatic dyslipidemia of atherogenic mice has rarely been studied. The specific aims of current study included to clarify lipidomic perturbation in liver tissues of apolipoprotein
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service