Skip to Content
Merck
All Photos(2)

Key Documents

850156P

Avanti

18:1 PI(3,4,5)P3

1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-[phosphoinositol-3,4,5-trisphosphate] (ammonium salt); PIP3[3′,4′,5′](18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H98N4O22P4
CAS Number:
Molecular Weight:
1171.17
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850156P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850156P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850156P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](OP(O)([O-])=O)[C@@H](OP(O)([O-])=O)[C@H]1O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+].[NH4+].[NH4+]

InChI key

CTYBALUTLRWYAN-CJXCTDJBSA-N

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Phosphatidylinositol is a structural phospholipid.[1] Phosphatidylinositol phosphate (PIP) is located in the plasma membrane.[2]

Application

18:1 PI(3,4,5)P3 is suitable to use in lipid stock preparation to form mixed micelles.[3] It has also been used:
  • in the preparation of planar-supported lipid bilayers (SLBs)[4]
  • in supplementing or enriching phagocyte membrane[5]
  • in liposome preparation[6]

Biochem/physiol Actions

Phosphoinositol-containing sphingolipids can induce ATPase at lower levels when compared to glycerophospholipids.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850156P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850156P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein kinase Czeta exhibits constitutive phosphorylation and phosphatidylinositol-3, 4, 5-triphosphate-independent regulation
Tobias IS, et al.
The Biochemical Journal, 473(4), 509-523 (2016)
Achieving a graded immune response: BTK adopts a range of active/inactive conformations dictated by multiple interdomain contacts
Joseph RE, et al.
Structure, 25(10), 1481-1494 (2017)
Phosphatidylinositol Turnover and Receptors
Ramazzotti G, et al.
Archives of Biochemistry and Biophysics (2016)
Mart G F Last et al.
ACS nano, 14(4), 4487-4498 (2020-04-03)
Membraneless organelles formed by liquid-liquid phase separation are dynamic structures that are employed by cells to spatiotemporally regulate their interior. Indeed, complex coacervation-based phase separation is involved in a multitude of biological tasks ranging from photosynthesis to cell division to
Rays H Y Jiang et al.
Cell host & microbe, 11(2), 99-100 (2012-02-22)
Survival of blood stage malaria parasites requires extensive host cell remodeling, which is facilitated by secretion of parasite proteins via a dedicated protein export pathway. In a recent Cell paper, Bhattacharjee et al., (2012) describe PI(3)P binding as one of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service