Skip to Content
Merck
All Photos(2)

Key Documents

840521P

Avanti

18:1 Lysyl PG

1,2-dioleoyl-sn-glycero-3-[phospho-rac-(3-lysyl(1-glycerol))] (chloride salt), powder

Synonym(s):

Lysyl-DOPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H93Cl2N2O11P
CAS Number:
Molecular Weight:
976.14
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (up to 15% acyl migration on lysine, TLC)

form

powder

packaging

pkg of 1 × 1 mg (840521P-1mg)
pkg of 1 × 10 mg (840521P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840521P

lipid type

phospholipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(COC([C@@](CCCC[NH3+])([H])[NH3+])=O)COP(O)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O.[Cl-].[Cl-]

InChI

1S/C48H91N2O11P.2ClH/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36-46(52)57-41-44(42-60-62(55,56)59-40-43(51)39-58-48(54)45(50)35-33-34-38-49)61-47(53)37-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;;/h17-20,43-45,51H,3-16,21-42,49-50H2,1-2H3,(H,55,56);2*

InChI key

QLNZUMYQGKEYKU-CIZZXKMDSA-N

General description

18:1 Lysyl PG/1,2-dioleoyl-sn-glycero-3-[phospho-rac-(3-lysyl(1-glycerol))] (lysyl-DOPG) is a synthetic phospholipid. It has a lysine group, added to the terminal glycerol of DOPG. Lysyl-phosphatidylglycerol (LPG), a membrane lipid is mainly seen in gram-positive bacteria. It is cationic phospholipid.

Application

18:1 Lysyl PG is suitable to prepare liposome solutions.

Biochem/physiol Actions

Lysyl-phosphatidylglycerol (lysyl-PG) in the cytoplasmic membranes of Staphylococcus is associated with improved resistance to antibiotics, like daptomycin and host defense antimicrobial peptides.

Packaging

5 mL Amber Glass Screw Cap Vial (840521P-1mg)
5 mL Clear Glass Sealed Ampule (840521P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysyl-phosphatidylglycerol attenuates membrane perturbation rather than surface association of the cationic antimicrobial peptide 6W-RP-1 in a model membrane system: implications for daptomycin resistance
Kilelee E, et al.
Antimicrobial Agents and Chemotherapy, 54(10), 4476-4479 (2010)
The lipid lysyl-phosphatidylglycerol is present in membranes of Rhizobium tropici CIAT899 and confers increased resistance to polymyxin B under acidic growth conditions
Sohlenkamp C, et al.
Molecular plant-microbe interactions : MPMI, 20(11), 1421-1430 (2007)
Helix stabilization precedes aqueous and bilayer-catalyzed fiber formation in islet amyloid polypeptide
Williamson JA, et al.
Journal of molecular biology, 393(2), 383-396 (2009)
Christian Sohlenkamp et al.
Molecular plant-microbe interactions : MPMI, 20(11), 1421-1430 (2007-11-06)
Lysyl-phosphatidylglycerol (LPG) is a well-known membrane lipid in several gram-positive bacteria but is almost unheard of in gram-negative bacteria. In Staphylococcus aureus, the gene product of mprF is responsible for LPG formation. Low pH-inducible genes, termed IpiA, have been identified
Lipid bilayer modules as determinants of K+ channel gating
Syeda R, et al.
Test, 289(7), 4233-4243 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service