Skip to Content
Merck
All Photos(3)

Documents

840072P

Avanti

Soy Lyso PC

Avanti Research - A Croda Brand 840072P, powder

Synonym(s):

L-α-lysophosphatidylcholine (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 2 × 100 mg (840072P-200mg)
pkg of 1 × 25 mg (840072P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840072P

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1

InChI key

ASWBNKHCZGQVJV-HSZRJFAPSA-N

General description

Lyso Phosphatidylcholine (LPC) or lysolecithin is a minor phospholipid in plasma (8-12%) and cellular membranes (<3%).

Application

Soy Lyso PC has been used in the synthesis of 15-methylhexadecanoic acid (15-MHDA) and glycerophospholipids.

Biochem/physiol Actions

Soy Lyso phosphocholine or phosphotidylcholine (PC) is among the other lipids present in soybean lecithin. Lysophosphatidylcholine (LPC) is a bioactive proinflammatory lipid. It is generated during pathological activities and is implicated in atherosclerosis and inflammatory diseases. LPC modifies the functions of numerous cell-types, including smooth muscle cells, endothelial cells, macrophages, monocytes, and T-cells. LPC activates several second messengers and is involved in signal transduction mechanisms.

Packaging

5 mL Clear Glass Sealed Ampule (840072P-200mg)
5 mL Clear Glass Sealed Ampule (840072P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine (lysolecithin) and its synthetic analogues. Immunemodulating and other biologic effects
Munder, PG and Modolell, M and Andreesen, Reinhard and Weltzien, HU and Westphal, O
Seminars in Immunopathology, 2(2), 187-203 (1979)
The use of natural and synthetic phospholipids as pharmaceutical excipients
van Hoogevest P and Wendel A
European Journal of Lipid Science and Technology, 116(9), 1088-1107 (2014)
High-Throughput Plasma Lipidomics: Detailed Mapping of the Associations with Cardiometabolic Risk Factors
Huynh K, et al.
Cell Chemical Biology, 26(1), 71-84 (2019)
Thomas Linemann et al.
Pharmaceutics, 5(2), 246-260 (2013-12-05)
The aim of the present study was to evaluate the transfection potential of chitosan-coated, green-fluorescent magnetic nanoparticles (MNPs) (chi-MNPs) after encapsulation inside polyethylglycol (PEG)ylated liposomes that produced lipid-encapsulated chitosan-coated MNPs (lip-MNPs), and also to evaluate how these particles would distribute
Role of lysophosphatidylcholine (LPC) in atherosclerosis
Matsumoto T, et al.
Current Medicinal Chemistry, 14(30), 3209-3220 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service