810166P
Avanti
18:1-12:0 NBD PG
Avanti Research™ - A Croda Brand 810166P, powder
Synonym(s):
1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C42H74N5O13P
CAS Number:
Molecular Weight:
888.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (810166P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 810166P
shipped in
dry ice
storage temp.
−20°C
General description
Phosphatidylglycerol (PG) is an anionic phospholipid. Phosphatidylglycerol (PG) is mostly found in the pulmonary surfactant and localized to the alveoli.(6) Nitrobenzoxadiazole phosphoglycerol (NBD PG) is a fluorescent analog with the fluorophore, 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) linked to the fatty acid chain of the phosphatidylglycerol.
Application
18:1-12:0 NBD PG might be used to study the use of fluorescence as a means of tracking the gold nanoparticles (GNPs) in live cells.
Biochem/physiol Actions
Phosphatidylglycerol (PG) is involved in the inflammatory mechanism and offers protection against viral infection.(6) It also contributes to the photosynthetic activity by maintaining the structural photosynthetic apparatus.
Packaging
5 mLAmber Glass Screw Cap Vial (810166P-1mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
Lipid-coated gold nanoparticles promote lamellar body formation in A549 cells
MeijingW and Nils OP
Biochimica et biophysica acta. General subjects null
Tandem mass spectrometry of novel ether-linked phospholipid analogs of anionic pulmonary surfactant phospholipids
Fickes R, et al.
Rapid Communications in Mass Spectrometry null
Direct Evidence for Requirement of Phosphatidylglycerol in
Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology null
Specificity of the transport of lipid II by FtsW in Escherichia coli
Mohammadi T
Journal of biological and chemical chronicles null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service