Skip to Content
Merck
All Photos(1)

Key Documents

800855P

Avanti

1,3-18:0 D5 DG

Avanti Research - A Croda Brand, powder

Synonym(s):

1,3-Dioctadecanoyl-2-hydroxy-sn-glycerol-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H71D5O5
CAS Number:
Molecular Weight:
630.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

form

powder

packaging

pkg of 1 × 1 mg (800855P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCCCCCC)=O)(O)C([2H])([2H])OC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3/i35D2,36D2,37D

InChI key

IZHVBANLECCAGF-RYBIQIASSA-N

Application

1,3-18:0 D5 DG has been used for the preparation of liposomes. It has also been used as an internal standard for lipid extraction.

Packaging

5 mL Amber Glass Screw Cap Vial (800855P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The exchangeable apolipoprotein Nplp2 sustains lipid flow and heat acclimation in Drosophila
Rommelaere S, et al.
Cell Reports, 27(3), 886-899 (2019)
Lipidomics analysis of behavioral variant frontotemporal dementia: a scope for biomarker development
Kim W, et al.
Frontiers in Neurology, 9(3), 104-104 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service