Skip to Content
Merck
All Photos(1)

Key Documents

791638C

Avanti

15:0-18:1-d7-PE

Avanti Research - A Croda Brand

Synonym(s):

1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H67D7NO8P
CAS Number:
Molecular Weight:
711.01
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791638C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (791638C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

15:0-18:1-d7-PE (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphoethanolamine) is a deuterated phospholipid. The lipid comprises phosphatidylethanolamine attached to pentadecanoic acid and oleic acid. Oleic acid harbors the deuterium ions. Phosphatidylethanolamine is a membrane phospholipid and serves as an intermediate in other glycerophospholipid synthesis.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7-PE (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphoethanolamine) has been used:
  • as a component of extraction solvent for lipid isolation from biological samples,
  • as an internal standard for spiking serum samples for liquid chromatography with tandem mass spectrometry (LC-MS/MS)
  • as an internal standard to spike plasma samples for fourier transform mass spectrometry (FTMS) analysis

Biochem/physiol Actions

Phosphatidylethanolamine (PE) regulates protein folding, protein modification, autophagy and cell division. PE serves as a substrate for glycosylphosphatidylinositol anchored proteins (GPI-APs) synthesis. Low levels of PE may be observed in Parkinson′s disease and may also inhibit protein maturation.

Packaging

2 mL Amber Glass Sealed Ampule (791638C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra F Gallego et al.
Biomolecules, 9(1) (2018-12-29)
Total fatty acid analysis is a routine method in many areas, including lipotyping of individuals in personalized medicine, analysis of foodstuffs, and optimization of oil production in biotechnology. This analysis is commonly done by converting fatty acyl (FA) chains of
Bjoern Titz et al.
Computational and structural biotechnology journal, 18, 1056-1073 (2020-05-19)
Cigarette smoke (CS) causes adverse health effects and, for smoker who do not quit, modified risk tobacco products (MRTPs) can be an alternative to reduce the risk of developing smoking-related diseases. Standard toxicological endpoints can lack sensitivity, with systems toxicology
Michael S Gardner et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1118-1119, 137-147 (2019-04-30)
Progress toward better diagnosis and treatment of lipid metabolism-related diseases requires high throughput approaches for multiplexed quantitative analysis of structurally diverse lipids, including phospholipids (PLs). This work demonstrates a simplified "one-pot" phospholipid extraction protocol, as an alternative to conventional liquid-liquid
Chuchu Wang et al.
Nature communications, 11(1), 1531-1531 (2020-03-27)
Vesicle associated membrane protein 2 (VAMP2/synaptobrevin2), a core SNARE protein residing on synaptic vesicles (SVs), forms helix bundles with syntaxin-1 and SNAP25 for the SNARE assembly. Prior to the SNARE assembly, the structure of VAMP2 is unclear. Here, by using
Dhaval Patel et al.
Oxidative medicine and cellular longevity, 2017, 4829180-4829180 (2017-08-09)
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service