Key Documents
Z569631
KDS syringe multi-rack for Legato series pumps
large
Synonym(s):
dosing pump syringe rack, infusion pump syringe rack
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
KD Scientific 78-8301
General description
Accommodates up to 4 x 60 to 140 mL syringes. Works with Legato 200, 210 and 210P.
Legal Information
Legato is a trademark of KD Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)