Skip to Content
Merck
All Photos(1)

Documents

W332518

Sigma-Aldrich

2,4,5-Trimethylthiazole

≥98%, FG

Synonym(s):

2,4,5-trimethyl-1,3-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NS
CAS Number:
Molecular Weight:
127.21
FEMA Number:
3325
EC Number:
Council of Europe no.:
11650
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.019
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.509 (lit.)

bp

166-167 °C/717 mmHg (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cocoa; coffee; musty; vegetable; nutty

SMILES string

Cc1nc(C)c(C)s1

InChI

1S/C6H9NS/c1-4-5(2)8-6(3)7-4/h1-3H3

InChI key

BAMPVSWRQZNDQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,4,5-Trimethylthiazole is a sulfur-containing heterocyclic compound mainly found in meat products such as chicken broth, cooked beef and roasted lamb fat.

Application


  • RIFM fragrance ingredient safety assessment, 2,4,5-trimethylthiazole, CAS Registry Number 13623-11-5.: A comprehensive safety assessment of 2,4,5-Trimethylthiazole used in fragrances, evaluating its impact on human health and the environment, underscoring its relevance in food and cosmetic industries (Api et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Primary odorants of chicken broth.
Gasser U & Grosch W.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 190(1), 3-8 (1990)
Heterocyclic compounds found in cooked meats.
Shibamoto T.
Journal of Agricultural and Food Chemistry, 28(2), 237-243 (1980)
Characterization of some volatile constituents of dry red beans.
Buttery RG, et al.
Journal of Agricultural and Food Chemistry, 23(3), 516-519 (1975)
Emmanuel Galliot et al.
Behavioural processes, 90(2), 161-166 (2012-01-18)
Odors can have repulsive effects on rodents based on two complementary adaptive behaviors: the avoidance of predator odors (potentially dangerous) and the avoidance of trigeminal stimulants (potentially noxious). The present study aimed to compare the behavioral effects on mice of
Suk-Woo Cho et al.
Biochemical and biophysical research communications, 325(2), 525-531 (2004-11-09)
Cyclic nucleotide-gated (CNG) channels encoded by tax-4 and tax-2 genes are required for chemo- and thermo-sensation in Caenorhabditis elegans. Here we report the identification and the characterization of cng-3, a new CNG channel gene, found in C. elegans. CNG-3 contains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service