Key Documents
Z568716
Sigma-Aldrich
Aldrich® combustion apparatus black paper sample wrappers
sample L × W 32 mm × 30 mm, pkg of 100 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pkg of 100 ea
Related Categories
General description
Absorb heat quickly for infrared ignition. Sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)