Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
Products
Product Category
spatulas/scoops (7)
Color
Manufacturer
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z560030
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 210 mm, blue, standard
Expand
View Pricing
Z561762
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 140 mm, white, anti-static, micro
Expand
View Pricing
Z560049
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 210 mm, white, standard
Expand
View Pricing
Z740505
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 140 mm, white, Biodegradable EcoSpatula
®
, short
Expand
View Pricing
Z560057
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 310 mm, green, macro
Expand
View Pricing
Z677787
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 210 mm, white, standard
Expand
View Pricing
Z742387
Levgo
®
smartSpatula
®
disposable PP laboratory spatula, Look for the V at the neck
™
,
L 210 mm, white, standard, sterile (EtO), endotoxin free (non-pyrogenic)
Expand
View Pricing