Skip to Content
Merck
All Photos(1)

Key Documents

W527602

Sigma-Aldrich

L-Isoleucine

99%, FCC, FG

Synonym(s):

(2S,3S)-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721792
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
odorless
grade:
FG
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.320

Assay

99%

form

crystalline powder

optical activity

[α]20/D +38.6 to +42.5°, c = 4 in 6 M HCl

mp

288 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CC[C@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Isoleucine is an essential amino acid used as a supplement in sports nutrition.[1] It is one of the free-amino acids found in common buckwheat.[2]

Application


  • Enhanced protein-metabolite correlation analysis: To investigate the association between Staphylococcus aureus mastitis and metabolic immune pathways.: This article discusses the metabolic pathways associated with bacterial infections in livestock, potentially useful for chemists studying metabolic pathways and their linkage to immune responses (Chen et al., 2024).

  • Probing pharmaceutically important amino acids L-isoleucine and L-tyrosine Solubilities: Unraveling the solvation thermodynamics in diverse mixed solvent systems.: This study explores the solubility and solvation thermodynamics of l-isoleucine, providing critical data for chemists working with pharmaceutical formulations and solubility studies (Chakraborty et al., 2024).


Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zorn H and Czermak P
Biotechnology of Food and Feed Additives., 219-220 null
Flavor components in buckwheat bread
Lin LY, et al
Journal of Food Processing and Preservation, 33(6), 814-826 (2009)
Urho M Kujala et al.
Circulation, 127(3), 340-348 (2012-12-22)
Long-term physical inactivity seems to cause many health problems. We studied whether persistent physical activity compared with inactivity has a global effect on serum metabolome toward reduced cardiometabolic disease risk. Sixteen same-sex twin pairs (mean age, 60 years) were selected
Eva Blomstrand et al.
The Journal of nutrition, 136(1 Suppl), 269S-273S (2005-12-21)
BCAAs (leucine, isoleucine, and valine), particularly leucine, have anabolic effects on protein metabolism by increasing the rate of protein synthesis and decreasing the rate of protein degradation in resting human muscle. Also, during recovery from endurance exercise, BCAAs were found
L Edvinsson
Blood vessels, 28(1-3), 35-45 (1991-01-01)
The cerebral circulation is supplied with two vasodilator systems: the parasympathetic system storing vasoactive intestinal peptide, peptide histidine isoleucine, acetylcholine and in a subpopulation of nerves neuropeptide Y, and the sensory system, mainly originating in the trigeminal ganglion, storing substance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service