Merck
  • Branched-chain amino acids activate key enzymes in protein synthesis after physical exercise.
Branched-chain amino acids activate key enzymes in protein synthesis after physical exercise.
The Journal of nutrition (2005-12-21)
Eva Blomstrand, Jörgen Eliasson, Håkan K R Karlsson, Rickard Köhnke
ABSTRACT
BCAAs (leucine, isoleucine, and valine), particularly leucine, have anabolic effects on protein metabolism by increasing the rate of protein synthesis and decreasing the rate of protein degradation in resting human muscle. Also, during recovery from endurance exercise, BCAAs were found to have anabolic effects in human muscle. These effects are likely to be mediated through changes in signaling pathways controlling protein synthesis. This involves phosphorylation of the mammalian target of rapamycin (mTOR) and sequential activation of 70-kD S6 protein kinase (p70 S6 kinase) and the eukaryotic initiation factor 4E-binding protein 1. Activation of p70 S6 kinase, and subsequent phopsphorylation of the ribosomal protein S6, is associated with enhanced translation of specific mRNAs. When BCAAs were supplied to subjects during and after one session of quadriceps muscle resistance exercise, an increase in mTOR, p70 S6 kinase, and S6 phosphorylation was found in the recovery period after the exercise with no effect of BCAAs on Akt or glycogen synthase kinase 3 (GSK-3) phosphorylation. Exercise without BCAA intake led to a partial phosphorylation of p70 S6 kinase without activating the enzyme, a decrease in Akt phosphorylation, and no change in GSK-3. It has previously been shown that leucine infusion increases p70 S6 kinase phosphorylation in an Akt-independent manner in resting subjects; however, a relation between mTOR and p70 S6 kinase has not been reported previously. The results suggest that BCAAs activate mTOR and p70 S6 kinase in human muscle in the recovery period after exercise and that GSK-3 is not involved in the anabolic action of BCAAs on human muscle.
Product Description
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Pricing and availability is not currently available.
SAFC
L-Isoleucine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.