Skip to Content
Merck
All Photos(1)

Key Documents

W396001

Sigma-Aldrich

Phenyl salicylate

≥99%, FG

Synonym(s):

Salol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
FEMA Number:
3960
Beilstein:
393969
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.689
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥99%

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; sweet

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-sensitivity between resorcinol, resorcinol monobenzoate and phenyl salicylate.
M Nakagawa et al.
Contact dermatitis, 27(3), 199-200 (1992-09-01)
P T Deota et al.
Natural product research, 17(1), 21-26 (2003-04-04)
The effect of photostabilization of azadirachtin-A (Aza-A) was examined in solutions when exposed to UV radiation, in the presence of four structurally different UV absorbers namely, p-aminobenzoic acid, 2,4-dihydroxybenzophenone, 4,4'-dihydroxybenzophenone and phenyl salicylate. The percentages of Aza-A recovered from the
C D Calnan et al.
Contact dermatitis, 7(4), 208-211 (1981-07-01)
Six cases of contact dermatitis from a lip salve are described. Five were allergic to phenyl salicylate and one to geraniol in the fragrance. The dermatitis spread in a ring around the mouth. Phenyl salicylate has been removed from the
J Baran et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 81(3 Pt 1), 031503-031503 (2010-04-07)
We report the investigation of glass-forming salol upon different courses of the temperature changes from liquid to glass state and back using FT-IR spectroscopy measurements in the wide spectral and temperature ranges. The formation of the ordered clusters in supercooled
M Niyaz Khan
Advances in colloid and interface science, 159(2), 160-179 (2010-08-03)
A new method, based upon semi-empirical kinetic approach, for the determination of ion exchange constant for ion exchange processes occurring between counterions at the cationic micellar surface is described in this review article. Basically, the method involves a reaction kinetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service