Skip to Content
Merck
All Photos(1)

Documents

W381101

Sigma-Aldrich

Neohesperidin dihydrochalcone

≥96%, FG

Synonym(s):

1-(4-((2-O-[6-Deoxy-α-L-mannopyranosyl]-β-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone, NHDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36O15
CAS Number:
Molecular Weight:
612.58
FEMA Number:
3811
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.061
E Number:
E 959
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥96%

mp

156-158 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

odorless

storage temp.

2-8°C

SMILES string

COc1ccc(CCC(=O)c2c(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc2O)cc1O

InChI

1S/C28H36O15/c1-11-21(34)23(36)25(38)27(40-11)43-26-24(37)22(35)19(10-29)42-28(26)41-13-8-16(32)20(17(33)9-13)14(30)5-3-12-4-6-18(39-2)15(31)7-12/h4,6-9,11,19,21-29,31-38H,3,5,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1

InChI key

ITVGXXMINPYUHD-CUVHLRMHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Neohesperidin dihydrochalcone is mainly used as a sweetener and flavoring agent. It can be prepared by the hydrogenation of neohesperidin, a bitter flavonone found in citrus fruits.

Application

  • Neohesperidin dihydrochalcone down-regulates MyD88-dependent and -independent signaling by inhibiting endotoxin-induced trafficking of TLR4 to lipid rafts.: This study explores the molecular mechanisms behind the immunomodulatory effects of Neohesperidin dihydrochalcone, revealing its potential to mitigate endotoxin-induced inflammatory responses (Xia et al., 2015).
  • Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Acute Tox. 4 Oral

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable

    Personal Protective Equipment

    dust mask type N95 (US), Eyeshields, Gloves

    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Alternative Sweeteners, 87-104 null
    Neohesperidin dihydrochalcone; an updated review on a naturally derived sweetener and flavor potentiator.
    Pratter PJ
    Perfumer & Flavorist (1980)
    J H Kroeze
    Chemical senses, 25(5), 555-559 (2000-10-04)
    Neohesperidin dihydrochalcone (NHDC) is an intensive sweetener, obtained by alkaline hydrogenation of neohesperidin. In this investigation a supposed taste enhancing effect of this substance was tested. A three-step procedure was used. In the first experiment, using a pool of 31
    R K Sahu et al.
    Mutation research, 89(1), 69-74 (1981-05-01)
    4 important flavonoid compounds, namely quercetin, kaempferol, neohesperidin dihydrochalcone and rutin, were administered to male mice for the detection of gross chromosomal anomalies by the micronucleus test. The first 3 compounds were positively clastogenic to different extents, whereas the 4th
    An exponential model for adaptation in taste.
    J F Gent
    Sensory processes, 3(4), 303-316 (1979-12-01)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service