Skip to Content
Merck
All Photos(3)

Documents

G2137

Sigma-Aldrich

Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice)

≥70% (HPLC)

Synonym(s):

3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid ammonium salt, Glycyrrhizin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
Beilstein:
3897076
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥70% (HPLC)

storage temp.

2-8°C

SMILES string

N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(O)=O)O[C@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)C(O)=O)C(O)=O

InChI

1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;/m0./s1

InChI key

ILRKKHJEINIICQ-OOFFSTKBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycyrrhizic acid (GA) is a non-selective inhibitor of 11β-hydroxysteroid dehydrogenase (11β-HSD), which mediates the inter-conversion of active and inactive glucocorticoids. GA also shows inhibitory effect on the cytokine activity of high-mobility group box 1 protein (HMGB1).

Application

The product has been used a source of glycyrrhizic acid (GA), to study its effect on glucose intolerance and insulin resistance in rats fed on high-fat/high-sucrose (HF/HS) diet.

Biochem/physiol Actions

Biologically active constituent in the sweet root of Glycyrrhiza species (licorice). Antiviral.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel inhibitory effects of Glycyrrhizic Acid on the accumulation of advanced glycation end product and its receptor expression.
Cheng HS, et al.
Natural products and bioprospecting, 4(6), 325-333 (2014)
Ashraf M Morgan et al.
Environmental science and pollution research international, 26(34), 34924-34930 (2019-10-30)
Atrazine (ATR) is a common herbicide used worldwide. It is a potent endocrine disruptor that causes hormonal imbalance. We investigated the modulatory role predisposed by glycyrrhizic acid (GA) against the hazardous effects caused by the ATR in the rabbit spleen.
High-mobility group box-1 was released actively and involved in LPS induced depressive-like behavior.
Wu TY, et al.
Journal of Psychiatric Research, 64, 99-106 (2015)
Hong Fan et al.
Journal of neuroinflammation, 17(1), 295-295 (2020-10-11)
Spinal cord injury (SCI) favors a persistent pro-inflammatory macrophages/microglia-mediated response with only a transient appearance of anti-inflammatory phenotype of immune cells. However, the mechanisms controlling this special sterile inflammation after SCI are still not fully elucidated. It is known that
Yu Okuma et al.
Neuropharmacology, 85, 18-26 (2014-05-27)
Glycyrrhizin (GL) is a major constituent of licorice root and has been suggested to inhibit the release of high mobility group box-1 (HMGB1), a protein considered representative of damage-associated molecular patterns. We found that GL bound HMGB1 but not RAGE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service