W363501
4-Hydroxy-5-methyl-3-furanone
97%, FG
Synonym(s):
toffee furanone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4OH2OCH3OH
CAS Number:
Molecular Weight:
114.10
FEMA Number:
3635
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.085
NACRES:
NA.21
Organoleptic:
caramel; coffee; brown; sweet
grade:
FG
Halal
Kosher
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
reg. compliance
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110
Assay
97%
mp
129-133 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
caramel; coffee; brown; sweet
SMILES string
CC1=C(O)C(=O)CO1
InChI
1S/C5H6O3/c1-3-5(7)4(6)2-8-3/h7H,2H2,1H3
InChI key
DLVYTANECMRFGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Hydroxy-5-methyl-3-furanone is the key flavor component of beef broth and shoyu (soy sauce).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Components contributing to beef flavor. Isolation of 4-hydroxy-5-methyl-3 (2H)-furanone and its 2, 5-dimethyl homolog from beef broth
Tonsbeek CH, et al.
Journal of Agricultural and Food Chemistry, 16(6), 1016-1021 (1968)
Mechanism of formation of 4-hydroxy-5-methyl-3 (2H)-furanone, a component of beef flavor, from Amadori products.
Hicks KB & Feather MS.
Journal of Agricultural and Food Chemistry, 23(5), 957-960 (1975)
Isolation of 4-hydroxy-5-methyl-3 (2H)-furanone, a flavor component in shoyu (soy sauce).
Nunomura N, et al.
Agricultural and Biological Chemistry, 43(6), 1361-1363 (1979)
Production of 4-hydroxy-5-methyl-3 (2H)-furanone, a component of beef flavor, from a 1-amino-1-deoxy-D-fructuronic acid
Hicks KB, et al.
Journal of Agricultural and Food Chemistry, 22(4), 724-725 (1974)
H J Kim et al.
Advances in experimental medicine and biology, 434, 91-99 (1998-05-23)
The chemometric principle was used to derive a guideline for obtaining a simple "yes or no" answer about the sterility of food particulates heated at aseptic processing temperatures. A quadratic temperature pulse model was used to estimate bacterial destruction from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service