Skip to Content
Merck
All Photos(1)

Key Documents

W352101

Sigma-Aldrich

Propyl mercaptan

≥97%

Synonym(s):

1-Propanethiol, n-Propylmercaptan, Mercaptan C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SH
CAS Number:
Molecular Weight:
76.16
FEMA Number:
3521
Beilstein:
1696860
EC Number:
Council of Europe no.:
11816
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.071
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 172.515

vapor density

2.54 (vs air)

vapor pressure

122 mmHg ( 20 °C)

Assay

≥97%

refractive index

n20/D 1.426 (lit.)

bp

67-68 °C (lit.)

mp

−113 °C (lit.)

density

0.820 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

onion; alliaceous

SMILES string

CCCS

InChI

1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3

InChI key

SUVIGLJNEAMWEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyl mercaptan is a volatile sulfur compound that is reported to occur in onion[1], potato[2] and wine.[3]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estimation of volatile sulphur compounds in beer.
Walker MD
Journal of the Institute of Brewing, 98(4), 283-287 (1992)
Food flavors and odors, volatile sulfur compounds in potatoes.
Gumbmann MR & Burr HK.
Journal of Agricultural and Food Chemistry, 12(5), 404-408 (1964)
Onion flavor and odor, the volatile flavor components of onions.
Carson JF & Wong FF
Journal of Agricultural and Food Chemistry, 9(2), 140-143 (1961)
Zeljko Sljivancanin et al.
Journal of the American Chemical Society, 124(49), 14789-14794 (2002-12-06)
Density functional theory calculations are carried out for the adsorption of a chiral molecule, (S)- and (R)-HSCH(2)CHNH(2)CH(2)P(CH(3))(2), on a chiral surface, Au(17 11 9)(S)(). The S-enantiomer is found to bind more strongly than the R-enantiomer by 8.8 kJ/mol, evidencing that
Noelia B Luque et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14514-14521 (2011-10-06)
Periodic density functional calculations have been carried out to investigate both the thiol adsorption on Au(111) surface and the reaction mechanism for the formation of the self-assembled monolayers, taking propanethiol as a representative example. The effect of coverage and surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service