Skip to Content
Merck
All Photos(1)

Key Documents

112925

Sigma-Aldrich

1-Butanethiol

99%

Synonym(s):

Butyl mercaptan, Mercaptan C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
Beilstein:
1730908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.1 (vs air)

Quality Level

vapor pressure

83 mmHg ( 37.7 °C)

Assay

99%

refractive index

n20/D 1.443 (lit.)

bp

98 °C (lit.)

mp

−116 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

functional group

thiol

SMILES string

CCCCS

InChI

1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

WQAQPCDUOCURKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butanethiol is used as a model gaseous analyte for fast-responding membrane-free amperometric gas sensor.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pratik Gurnani et al.
Macromolecular bioscience, 18(10), e1800213-e1800213 (2018-08-08)
Current approaches to generate core-shell nanoparticles for biomedical applications are limited by factors such as synthetic scalability and circulatory desorption of cytotoxic surfactants. Developments in controlled radical polymerization, particularly in dispersed states, represent a promising method of overcoming these challenges.
Yinghui Chen et al.
Acta biomaterialia, 102, 384-393 (2019-12-04)
Surface modification to obtain high dispersion stability and biocompatibility is a key factor for bio-application of upconversion nanoparticles (UCNPs). A systematic study of UCNPs modified with four hydrophilic molecules separately, comparing their dispersion stability in biological buffers and cellular biocompatibility
X Wang et al.
Chemical science, 9(21), 4879-4891 (2018-06-19)
Acidithiobacillus ferrooxidans, a chemolithoautotrophic Gram-negative bacterium, has a remarkable ability to obtain energy from ferrous iron oxidation at pH 2. Several metalloproteins have been described as being involved in this respiratory chain coupling iron oxidation with oxygen reduction. However, their
Late acute, delayed neurotoxic and cholinergic effects of S,S,S-tributyl phosphorotrithioite (Merphos) in hens.
M B Abou-Donia et al.
Toxicology and applied pharmacology, 53(3), 439-457 (1980-05-01)
Rosanna Toniolo et al.
Analytical and bioanalytical chemistry, 405(11), 3571-3577 (2012-12-13)
A prototype of a fast-response task-specific amperometric gas sensor based on paper-supported room-temperature ionic liquids (RTILs) is proposed here for improved analysis of volatile acid species. It consists of a small filter paper foil soaked with a RTIL mixture containing

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service